Skip to Content
Merck
All Photos(1)

Key Documents

EHU053371

Sigma-Aldrich

MISSION® esiRNA

targeting human SYP

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$335.00
50 μG
$584.00

$335.00


Check Cart for Availability


Select a Size

Change View
20 μG
$335.00
50 μG
$584.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

$335.00


Check Cart for Availability

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

ACCGAGAGTGACCTCAGCATCGAGGTCGAGTTCGAGTACCCCTTCAGGCTGCACCAAGTGTACTTTGATGCACCCACCTGCCGAGGGGGCACCACCAAGGTCTTCTTAGTTGGGGACTACTCCTCGTCAGCCGAATTCTTTGTCACCGTGGCCGTGTTTGCCTTCCTCTACTCCATGGGGGCTCTGGCCACCTACATCTTCCTGCAGAACAAGTACCGAGAGAATAACAAAGGGCCCATGCTGGACTTTCTGGCCACGGCTGTGTTCGCCTTCATGTGGCTAGTTAGCTCATCGGCATGGGCCAAGGGGCTGTCAGATGTGAAGATGGCCACAGACCCAGAGAACATTATCAAGGAGATGCCTGTCTGCCGCCAGACAGGGAACACATGCAAGGAGCTGA

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Teng-Fei Li et al.
Scientific reports, 7, 45056-45056 (2017-03-23)
Bulleyaconitine (BAA) has been shown to possess antinociceptive activities by stimulation of dynorphin A release from spinal microglia. This study investigated its underlying signal transduction mechanisms. The data showed that (1) BAA treatment induced phosphorylation of CREB (rather than NF-κB)
Desheng Zhong et al.
Journal of cellular biochemistry, 115(9), 1624-1635 (2014-05-03)
Pan-Bcl-2 family inhibitor obatoclax has been demonstrated to be effective against various cancers, of which the mechanism of action is not fully understood. In this study, we demonstrate that obatoclax suppressed esophageal cancer cell viability with concomitant G1/G0-phase cell cycle
Xi-Xi Lin et al.
Toxicology mechanisms and methods, 24(8), 575-583 (2014-08-20)
Cigarette smoke contains reactive oxygen (ROS) that can cause oxidative stress. It increases the number of apoptotic and necrotic lung cells and further induces the development of chronic airway disease. In this study, we investigated the effects of cigarette smoke
Xiaochun Yang et al.
Oncotarget, 6(8), 6203-6217 (2015-03-10)
Liver dysfunction is a common side effect associated with the treatment of dasatinib and its mechanism is poorly understood. Autophagy has been thought to be a potent survival or death factor for liver dysfunction, which may shed the light on
Bruce C McGorum et al.
Molecular & cellular proteomics : MCP, 14(11), 3072-3086 (2015-09-15)
Equine grass sickness (EGS) is an acute, predominantly fatal, multiple system neuropathy of grazing horses with reported incidence rates of ∼2%. An apparently identical disease occurs in multiple species, including but not limited to cats, dogs, and rabbits. Although the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service