Skip to Content
Merck
All Photos(1)

Key Documents

EHU050181

Sigma-Aldrich

MISSION® esiRNA

targeting human LRG1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CCATCTCCTGTCAACCACCTGCCGAAATCCCCGGCTACCTGCCAGCCGACACCGTGCACCTGGCCGTGGAATTCTTCAACCTGACCCACCTGCCAGCCAACCTCCTCCAGGGCGCCTCTAAGCTCCAAGAATTGCACCTCTCCAGCAATGGGCTGGAAAGCCTCTCGCCCGAATTCCTGCGGCCAGTGCCGCAGCTGAGGGTGCTGGATCTAACCCGAAACGCCCTGACCGGGCTGCCCCCGGGCCTCTTCCAGGCCTCAGCCACCCTGGACACCCTGGTATTGAAAGAAAACCAGCTGGAGGTCCTGGAGGTCTCGTGGCTACACGGCCTGAAAGCTCTGGGGCATCTGGACCTGTCTGGGAACCGCCTCCGGAAACTGCCCCCCGGGCTGCTGGCCAACTTCACCCTCCTGCGCACCCTTGACCTTGGGGAGAACCAGTTGGAGACCTTGC

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lan Luan et al.
Experimental and therapeutic medicine, 21(4), 367-367 (2021-03-19)
Retinoblastoma (RB) is the most common primary intraocular cancer type that occurs during retinal development in childhood. Previous studies have reported that long non-coding RNAs (lncRNAs) are involved in the development of RB. Therefore, the aim of the present study
Yiyun Wang et al.
Cell death & disease, 8(3), e2715-e2715 (2017-03-31)
The incomplete understanding of aberrant neovascularization, which contributes to osteoarthritis suggests that additional modulators have yet to be identified. Our objective was to identify the role of Leucine-rich-alpha-2-glycoprotein1 (LRG1), a new regulator of pathogenic angiogenesis, in osteoarthritis progression and to
Qian Zhang et al.
OncoTargets and therapy, 11, 2745-2752 (2018-05-23)
Leucine-rich α-2-glycoprotein-1 (LRG1) is differentially expressed in many kinds of diseases including cancer, however, it has not been thoroughly studied yet. The objective of this study was to detect the expression and potential mechanism of LRG1 in colorectal cancer (CRC).
Masaaki Yamamoto et al.
Cancer science, 108(10), 2052-2060 (2017-07-27)
Gastric cancer is one of the most common malignant tumors. Although improvement in chemotherapy has been achieved, the clinical prognosis of advanced gastric cancer remains poor. Therefore, it is increasingly important to predict the prognosis and determine whether patients should
Gu Gong et al.
Journal of molecular neuroscience : MN, 54(1), 20-26 (2014-02-15)
Lipopolysaccharide (LPS) preconditioning is a powerful neuroprotective phenomenon by which an injurious stimulus renders the brain resistant to a subsequent damaging ischemic insult. The LPS response gene (Lrg) is a recently identified gene in human dental pulp cells treated with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service