Skip to Content
Merck
All Photos(1)

Key Documents

EHU028441

Sigma-Aldrich

MISSION® esiRNA

targeting human STIM2

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$335.00
50 μG
$584.00

$335.00


Check Cart for Availability


Select a Size

Change View
20 μG
$335.00
50 μG
$584.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

$335.00


Check Cart for Availability

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TCCCTGCATGTCACTGAGTCCACCATGCTTTACAGAAGAAGACAGATTTAGTCTGGAAGCTCTTCAAACAATACATAAACAAATGGATGATGACAAAGATGGTGGAATTGAAGTAGAGGAAAGTGATGAATTCATCAGAGAAGATATGAAATATAAAGATGCTACTAATAAACACAGCCATCTGCACAGAGAAGATAAACATATAACGATTGAGGATTTATGGAAACGATGGAAAACATCAGAAGTTCATAATTGGACCCTTGAAGACACTCTTCAGTGGTTGATAGAGTTTGTTGAACTACCCCAATATGAGAAGAATTTTAGAGACAACAATGTCAAAGGAACGACACTTCCCAGGATAGCAGTGCACGAACCTTCATTTATGATCTCCCAGTTGAAAATCAGTGACCGGAGTCACAGACAAAAACTTCAGCTCAAGGCATTGGATGTGGTTTTGTTTGGACCTCTAACACGCCCACCTCAT

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jiwang Chen et al.
Circulation, 135(16), 1532-1546 (2017-02-17)
Pulmonary arterial hypertension is a severe and progressive disease, a hallmark of which is pulmonary vascular remodeling. Nicotinamide phosphoribosyltransferase (NAMPT) is a cytozyme that regulates intracellular nicotinamide adenine dinucleotide levels and cellular redox state, regulates histone deacetylases, promotes cell proliferation
Vladimir A Vigont et al.
Frontiers in cell and developmental biology, 9, 625231-625231 (2021-02-20)
Huntington's disease (HD) is a severe autosomal-dominant neurodegenerative disorder caused by a mutation within a gene, encoding huntingtin protein. Here we have used the induced pluripotent stem cell technology to produce patient-specific terminally differentiated GABA-ergic medium spiny neurons modeling a
Raz Palty et al.
Cell research, 25(8), 963-980 (2015-07-04)
Calcium flux through store-operated calcium entry is a major regulator of intracellular calcium homeostasis and various calcium signaling pathways. Two key components of the store-operated calcium release-activated calcium channel are the Ca(2+)-sensing protein stromal interaction molecule 1 (STIM1) and the
Jingsheng Xia et al.
The Journal of physiology, 592(16), 3443-3461 (2014-05-27)
Store-operated calcium channels (SOCs) are calcium-selective cation channels that mediate calcium entry in many different cell types. Store-operated calcium entry (SOCE) is involved in various cellular functions. Increasing evidence suggests that impairment of SOCE is responsible for numerous disorders. A
Ruby A Fernandez et al.
American journal of physiology. Cell physiology, 308(8), C581-C593 (2015-02-13)
Pulmonary arterial hypertension (PAH) is a progressive disease that, if left untreated, eventually leads to right heart failure and death. Elevated pulmonary arterial pressure (PAP) in patients with PAH is mainly caused by an increase in pulmonary vascular resistance (PVR).

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service