Skip to Content
Merck
All Photos(1)

Key Documents

EHU026581

Sigma-Aldrich

MISSION® esiRNA

targeting human TTK

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$335.00
50 μG
$584.00

$335.00


Check Cart for Availability


Select a Size

Change View
20 μG
$335.00
50 μG
$584.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

$335.00


Check Cart for Availability

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

AGGAAGTGGAGGTTCAAGCAAGGTATTTCAGGTGTTAAATGAAAAGAAACAGATATATGCTATAAAATATGTGAACTTAGAAGAAGCAGATAACCAAACTCTTGATAGTTACCGGAACGAAATAGCTTATTTGAATAAACTACAACAACACAGTGATAAGATCATCCGACTTTATGATTATGAAATCACGGACCAGTACATCTACATGGTAATGGAGTGTGGAAATATTGATCTTAATAGTTGGCTTAAAAAGAAAAAATCCATTGATCCATGGGAACGCAAGAGTTACTGGAAAAATATGTTAGAGGCAGTTCACACAATCCATCAACATGGCATTGTTCACAGTGATCTTAAACCAGCTAACTTTCTGATAGTTGATGGAATGCTAAAGCTAATTGATTTTGGGATTGCAAACCAA

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mi-Young Lee et al.
Cell division, 9, 3-3 (2014-10-04)
Centrosome amplification (CA) amongst particular breast cancer subtypes (Her2+ subtype) is associated with genomic instability and aggressive tumor phenotypes. However, changes in signaling pathways associated with centrosome biology have not been fully explored in subtype specific models. Novel centrosome regulatory
Xing Liu et al.
Oncotarget, 6(33), 34309-34320 (2015-09-30)
Hepatocellular carcinoma (HCC) is one of the most malignant cancers with poor clinical outcome. The protein kinase human monopolar spindle 1 (hMps1/TTK) gene expression is significantly increased in HCCs. However, its contributions to hepatocarcinogenesis remain unclear. In this study, we

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service