Skip to Content
Merck
All Photos(1)

Key Documents

EHU023531

Sigma-Aldrich

MISSION® esiRNA

targeting human RB1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TGCATGGCTCTCAGATTCACCTTTATTTGATCTTATTAAACAATCAAAGGACCGAGAAGGACCAACTGATCACCTTGAATCTGCTTGTCCTCTTAATCTTCCTCTCCAGAATAATCACACTGCAGCAGATATGTATCTTTCTCCTGTAAGATCTCCAAAGAAAAAAGGTTCAACTACGCGTGTAAATTCTACTGCAAATGCAGAGACACAAGCAACCTCAGCCTTCCAGACCCAGAAGCCATTGAAATCTACCTCTCTTTCACTGTTTTATAAAAAAGTGTATCGGCTAGCCTATCTCCGGCTAAATACACTTTGTGAACGCCTTCTGTCTGAGCACCCAGAATTAGAACATATCATCTGGACCCTTTTCCAGCACACCCTGCAGAATGAGTATGAACTCATGAGAGACAGGCATTTGGACC

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Toshiyuki Sumi et al.
Biochemical and biophysical research communications, 501(1), 253-258 (2018-05-05)
High expression levels of survivin in KRAS-mutant lung adenocarcinomas are linked with unfavorable patient outcomes, suggesting that survivin is a promising target for tumor treatment. We found that trametinib, a MEK inhibitor, downregulates survivin expression in the RB1-positive KRAS-mutant lung
Chun-Yu Liu et al.
PloS one, 12(12), e0189007-e0189007 (2017-12-21)
Triple negative breast cancer (TNBC) lacks specific drug targets and remains challenging. Palbociclib, a cyclin-dependent kinases 4 and 6 (CDK4/6) inhibitor is approved for metastatic estrogen receptor (ER)-positive and human epithermal growth factor 2 (HER2)-negative breast cancer. The nature of
Elisabetta Marangoni et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 24(11), 2605-2615 (2018-02-22)
Purpose: Triple-negative breast cancer (TNBC) patients with residual disease after neoadjuvant chemotherapy have a poor outcome. We developed patient-derived xenografts (PDX) from residual tumors to identify efficient chemotherapies and predictive biomarkers in a context of resistance to anthracyclines- and taxanes-based
Karen McColl et al.
Oncotarget, 8(43), 73745-73756 (2017-11-02)
The majority of small cell lung cancer (SCLC) patients demonstrate initial chemo-sensitivity, whereas a distinct subgroup of SCLC patients, termed chemo-refractory, do not respond to treatment. There is little understanding of how to distinguish these patients prior to disease treatment.
Gabrielle van Caloen et al.
Molecular cancer therapeutics, 19(3), 777-789 (2020-01-12)
Cell-cycle pathway impairments resulting in CDK4 and 6 activation are frequently observed in human papillomavirus (HPV)-negative squamous cell carcinoma of the head and neck (SCCHN). We investigated the activity of ribociclib, a CDK4/6 inhibitor, in SCCHN models with the aim

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service