Skip to Content
Merck
All Photos(1)

Key Documents

EHU020151

Sigma-Aldrich

MISSION® esiRNA

targeting human ADAM9

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$335.00
50 μG
$584.00

$335.00


Estimated to ship on11 April 2025



Select a Size

Change View
20 μG
$335.00
50 μG
$584.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

$335.00


Estimated to ship on11 April 2025


description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

AGGCGGGATTAATGTGTTTGGACAAATCACTGTGGAGACATTTGCTTCCATTGTTGCTCATGAATTGGGTCATAATCTTGGAATGAATCACGATGATGGGAGAGATTGTTCCTGTGGAGCAAAGAGCTGCATCATGAATTCAGGAGCATCGGGTTCCAGAAACTTTAGCAGTTGCAGTGCAGAGGACTTTGAGAAGTTAACTTTAAATAAAGGAGGAAACTGCCTTCTTAATATTCCAAAGCCTGATGAAGCCTATAGTGCTCCCTCCTGTGGTAATAAGTTGGTGGACGCTGGGGAAGAGTGTGACTGTGGTACTCCAAAGGAATGTGAATTGGACCCTTGCTGCGAAGGAAGTACCTGTAAGCTTAAATCATTTGCTGAGTGTGCATATGGTGACTGTTGTAAAGACTGTCGGTTCCTTCCA

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rui Zhou et al.
Frontiers in medicine, 7, 214-214 (2020-07-09)
Upregulation of a disintegrin and metalloprotease 9 (ADAM9) is correlated with progression of cancers, such as prostate, bladder, and pancreatic cancers. However, its role in triple-negative breast cancer (TNBC) is still unclear. Our study aimed to investigate whether ADAM9 is
Jun Arai et al.
Journal of gastroenterology and hepatology, 33(5), 1075-1081 (2017-10-22)
The multi-kinase inhibitor regorafenib (REG) was recently demonstrated to be effective in patients with sorafenib (SOR)-resistant hepatocellular carcinoma (HCC). Interestingly, SOR is known to enhance the accumulation of membrane-bound MHC class I polypeptide-related sequence A (mMICA) in HCC cells and
Mari Ueno et al.
Cancer science, 109(2), 471-482 (2017-12-17)
ADAMs (a disintegrin and metalloproteinases) are involved in various biological events such as cell adhesion, migration and invasion, membrane protein shedding and proteolysis. However, there have been no systematic studies on the expression of ADAMs in human ovarian carcinomas. We
Liang Chang et al.
Molecular medicine reports, 12(1), 1197-1204 (2015-03-18)
A disintegrin and metalloproteinase 9 (ADAM9) is a type I transmembrane protein that has been associated with cancer development and metastasis in various types of cancer. However, little is known about its role in non-small cell lung cancer (NSCLC). The
Jun Arai et al.
Cancer immunology, immunotherapy : CII, 70(1), 203-213 (2020-07-20)
In our previous genome-wide association study, we demonstrated the association between MHC class I-related chain A (MICA) and hepatocellular carcinoma (HCC) development in patients with chronic hepatitis C. Increasing membrane-bound MICA (mMICA) in cancer cells by reducing MICA sheddases facilitates

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service