Skip to Content
Merck
All Photos(1)

Key Documents

EHU014831

Sigma-Aldrich

MISSION® esiRNA

targeting human BUB1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GGACCCAGTTGATGGAAAGACTAAAGCCATCTATGCAGCACATGTTTATGAAGTTCTATTCTGCCCACTTATTCCAGAATGGCAGTGTATTAGTAGGAGAGCTCTACAGCTATGGAACATTATTAAATGCCATTAACCTCTATAAAAATACCCCTGAAAAAGTGATGCCTCAAGGTCTTGTCATCTCTTTTGCTATGAGAATGCTTTACATGATTGAGCAAGTGCATGACTGTGAAATCATTCATGGAGACATTAAACCAGACAATTTCATACTTGGAAACGGGCAAGTATTTTTGGAACAGGATGATGAAGATGATTTATCTGCTGGCTTGGCACTGATTGACCTGGGTCAGAGTATAGATATGAAACTTTTTCCAAAAGGAACTATATTCACAGCAAAGTGTGAAACATCTGGTTTTCAGTGTGTTGAGATGCTCAGCAACAAACCATGGAACTACCAGATCGATTACTTTGGGGTTGCTGCAACAGTATATTGCATGCTCTTTGGCAC

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

Related Categories

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mathijs Vleugel et al.
Journal of cell science, 128(16), 2975-2982 (2015-07-08)
Mitotic chromosome segregation is initiated by the anaphase promoting complex/cyclosome (APC/C) and its co-activator CDC20 (forming APC/C(CDC20)). APC/C(CDC20) is inhibited by the spindle assembly checkpoint (SAC) when chromosomes have not attached to spindle microtubules. Unattached kinetochores catalyze the formation of
Yutaka Matsubara et al.
Shock (Augusta, Ga.), 51(3), 364-371 (2018-04-03)
Severe sepsis is critical to health and can result in acute renal failure (ARF). Tissue factor (TF) and thrombomodulin (TM) play key roles in vascular endothelial functions by helping maintain microcirculation in the kidney. Budding uninhibited by benzimidazole-1 (Bub1) plays
Katharina Overlack et al.
Current biology : CB, 27(19), 2915-2927 (2017-09-26)
The spindle assembly checkpoint (SAC) prevents premature sister chromatid separation during mitosis. Phosphorylation of unattached kinetochores by the Mps1 kinase promotes recruitment of SAC machinery that catalyzes assembly of the SAC effector mitotic checkpoint complex (MCC). The SAC protein Bub3
Grégory Eot-Houllier et al.
Nature communications, 9(1), 1888-1888 (2018-05-16)
Sustained spindle tension applied to sister centromeres during mitosis eventually leads to uncoordinated loss of sister chromatid cohesion, a phenomenon known as "cohesion fatigue." We report that Aurora A-dependent phosphorylation of serine 7 of the centromere histone variant CENP-A (p-CENP-AS7)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service