Skip to Content
Merck
All Photos(1)

Key Documents

EHU008731

Sigma-Aldrich

MISSION® esiRNA

targeting human NR4A2

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$335.00
50 μG
$584.00

$335.00


Check Cart for Availability


Select a Size

Change View
20 μG
$335.00
50 μG
$584.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

$335.00


Check Cart for Availability

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CCCAGTGGAGGGTAAACTCATCTTTTGCAATGGGGTGGTCTTGCACAGGTTGCAATGCGTTCGTGGCTTTGGGGAATGGATTGATTCCATTGTTGAATTCTCCTCCAACTTGCAGAATATGAACATCGACATTTCTGCCTTCTCCTGCATTGCTGCCCTGGCTATGGTCACAGAGAGACACGGGCTCAAGGAACCCAAGAGAGTGGAAGAACTGCAAAACAAGATTGTAAATTGTCTCAAAGACCACGTGACTTTCAACAATGGGGGGTTGAACCGCCCCAATTATTTGTCCAAACTGTTGGGGAAGCTCCCAGAACTTCGTACCCTTTGCACACAGGGGCTACAGCGCATTTTCTACCTGAAATTGGAAGACTTGGTGCCACCGCCAGCAATAATTGACAAACTTTTCCTGGACACTTTACCTTTCTAAGACCTCCTCCCAAGCACTTCAAAGGAACTGGAATGATAATGGAAACTGTCAAGAGGGGGCAAGTCACATGGGCAGAGATAGCCGTGTGAGCAGTCTCAGCTC

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Soo Min Kim et al.
Molecules and cells, 43(6), 551-571 (2020-06-12)
Nuclear receptor-related 1 (Nurr1) protein has been identified as an obligatory transcription factor in midbrain dopaminergic neurogenesis, but the global set of human NURR1 target genes remains unexplored. Here, we identified direct gene targets of NURR1 by analyzing genome-wide differential
Shawn Llopis et al.
BMC cancer, 13, 139-139 (2013-03-23)
NR4A orphan nuclear receptors are involved in multiple biological processes which are important in tumorigenesis such as cell proliferation, apoptosis, differentiation, and glucose utilization. The significance of NR4A family member NURR1 (NR4A2) in breast cancer etiology has not been elucidated.
Xin Heng et al.
Molecular neurodegeneration, 7, 4-4 (2012-02-03)
NURR1 (also named as NR4A2) is a member of the steroid/thyroid hormone receptor family, which can bind to DNA and modulate expression of target genes. Previous studies have shown that NURR1 is essential for the nigral dopaminergic neuron phenotype and
Masanori Nagaoka et al.
Journal of immunology (Baltimore, Md. : 1950), 199(8), 2958-2967 (2017-09-13)
NR4A3/NOR1 belongs to the NR4A subfamily of the nuclear hormone receptor superfamily, which is activated in a ligand-independent manner. To examine the role of NR4A3 in gene expression of dendritic cells (DCs), we introduced NR4A3 small interfering RNA (siRNA) into
S Loppi et al.
Brain, behavior, and immunity, 73, 670-681 (2018-08-01)
Ischemic stroke is amongst the leading causes of death and disabilities. The available treatments are suitable for only a fraction of patients and thus novel therapies are urgently needed. Blockage of one of the cerebral arteries leads to massive and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service