Skip to Content
Merck
All Photos(1)

Key Documents

EHU007551

Sigma-Aldrich

MISSION® esiRNA

targeting human LCN2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CAATTCCAGGGGAAGTGGTATGTGGTAGGCCTGGCAGGGAATGCAATTCTCAGAGAAGACAAAGACCCGCAAAAGATGTATGCCACCATCTATGAGCTGAAAGAAGACAAGAGCTACAATGTCACCTCCGTCCTGTTTAGGAAAAAGAAGTGTGACTACTGGATCAGGACTTTTGTTCCAGGTTGCCAGCCCGGCGAGTTCACGCTGGGCAACATTAAGAGTTACCCTGGATTAACGAGTTACCTCGTCCGAGTGGTGAGCACCAACTACAACCAGCATGCTATGGTGTTCTTCAAGAAAGTTTCTCAAAACAGGGAGTACTTCAAGATCACCCTCTACGGGAGAACCAAGGAGCTGACTTCGGAACTAAAGGAGAACTTCATCCGCTTCTCCAAATCTCTGGGCCTCCCTGAAAACCACATCGTCTTCCCTGTC

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Se-Lim Kim et al.
Cancer science, 108(11), 2176-2186 (2017-09-01)
Lipocalin 2 (LCN2), a member of the lipocalin superfamily, plays an important role in oncogenesis and progression in various types of cancer. However, the expression pattern and functional role of LCN2 in colorectal cancer (CRC) is still poorly understood. The
Masafumi Okuda et al.
Oncotarget, 8(21), 34670-34677 (2017-04-15)
Esophageal cancer is the eighth most common cancer and the sixth most common cause of cancer-related deaths worldwide. Despite the research progress in understanding the disease, the mechanism underlying the metastasis is still unclear. Here, we successfully generated a highly
Bilge Ören et al.
The Journal of pathology, 239(3), 274-285 (2016-04-03)
Tumour cell-secreted factors skew infiltrating immune cells towards a tumour-supporting phenotype, expressing pro-tumourigenic mediators. However, the influence of lipocalin-2 (Lcn2) on the metastatic cascade in the tumour micro-environment is still not clearly defined. Here, we explored the role of stroma-derived
Ioanna Mosialou et al.
The Journal of experimental medicine, 217(10) (2020-07-09)
Regulation of food intake is a recently identified endocrine function of bone that is mediated by Lipocalin-2 (LCN2). Osteoblast-secreted LCN2 suppresses appetite and decreases fat mass while improving glucose metabolism. We now show that serum LCN2 levels correlate with insulin
Peng Guo et al.
Theranostics, 6(1), 1-13 (2016-01-02)
Lipocalin 2 (Lcn2) is a promising therapeutic target as well as a potential diagnostic biomarker for breast cancer. It has been previously shown to promote breast cancer progression by inducing the epithelial to mesenchymal transition in breast cancer cells as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service