Skip to Content
Merck
All Photos(1)

Key Documents

EHU006001

Sigma-Aldrich

MISSION® esiRNA

targeting human FANCF

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$335.00
50 μG
$584.00

$335.00


Check Cart for Availability


Select a Size

Change View
20 μG
$335.00
50 μG
$584.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

$335.00


Check Cart for Availability

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

ACCCAAATCTCCAGGAGGACTCTCTGATGAAGACCCAGGCGGAGCTGCTGCTGGAGCGTCTGCAGGAGGTGGGGAAGGCCGAAGCGGAGCGTCCCGCCAGGTTTCTCAGCAGCCTGTGGGAGCGCTTGCCTCAGAACAACTTCCTGAAGGTGATAGCGGTGGCGCTGTTGCAGCCGCCTTTGTCTCGTCGGCCCCAAGAAGAGTTGGAACCCGGCATCCACAAATCACCTGGAGAGGGGAGCCAAGTGCTAGTCCACTGGCTTCTGGGGAATTCGGAAGTCTTTGCTGCCTTTTGTCGCGCCCTCCCAGCCGGGCTTTTGACTTTAGTGACTAGCCGCCACCCAGCGCTGTCTCCTGTCTATCTGGGTCTGCTAACAGACTGGGGTCAACGTTTGCACTATGACCTTCAGAAAGGCATTTGGGTTGGAACTGAGTCC

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yanlin Li et al.
PloS one, 7(8), e44254-e44254 (2012-09-07)
Fanconi anemia complementation group-F (FANCF) is a key factor to maintain the function of FA/BRCA, a DNA-damage response pathway. However, the functional role of FANCF in breast cancer has not been elucidated. In this study, we examined the effects and
Miao He et al.
Oncology reports, 29(5), 1721-1729 (2013-02-27)
In the present study, we downregulated FANCF expression by small interfering RNA (siRNA) in OVCAR ovarian cancer cells to address the effects of decreased FANCF expression on the function of the Fanconi anemia (FA)/breast cancer susceptibility gene (BRCA) pathway. Furthermore
Jiankun Yu et al.
Journal of breast cancer, 16(3), 291-299 (2013-10-25)
Fanconi anemia complementation group F (FANCF) is a key factor to maintaining the function of Fanconi anaemia/BRCA (FA/BRCA) pathway, a DNA-damage response pathway. However, the functional role of FANCF in breast cancer has not been elucidated. In the present study
Jiankun Yu et al.
International journal of nanomedicine, 11, 6651-6666 (2016-12-21)
Two different disulfide (SS)-containing poly(amidoamine) (PAA) polymers were constructed using guanidino (Gua)-containing monomers (ie, arginine [Arg] and agmatine [Agm]) and N,N'-cystamine bisacrylamide (CBA) by Michael-addition polymerization. In order to characterize these two Gua-SS-PAA polymers and investigate their potentials as short
Lin Zhao et al.
International journal of oncology, 45(1), 129-138 (2014-05-03)
The Fanconi anemia/BRCA (FA/BRCA) DNA damage repair pathway plays a pivotal role in the cellular response to DNA alkylating agents and greatly influences drug response in cancer treatment. However, the molecular mechanisms underlying the FA/BRCA pathway reversed resistance have received

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service