Skip to Content
Merck
All Photos(1)

Key Documents

E9645

Sigma-Aldrich

Estrone sulfamate

≥98%

Synonym(s):

1,3,5(10)-Estratrien-17-one 3-sulfamate

Sign Into View Organizational & Contract Pricing

Select a Size

0.2 MG
$318.00

$318.00


Estimated to ship on17 May 2025



Select a Size

Change View
0.2 MG
$318.00

About This Item

Empirical Formula (Hill Notation):
C18H23NO4S
CAS Number:
Molecular Weight:
349.44
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77

$318.00


Estimated to ship on17 May 2025


biological source

synthetic (organic)

Quality Level

Assay

≥98%

form

powder

storage temp.

−20°C

SMILES string

[S](=O)(=O)(N)Oc1cc2c(cc1)[C@@H]3[C@H]([C@H]4[C@](CC3)(C(=O)CC4)C)CC2

InChI

1S/C18H23NO4S/c1-18-9-8-14-13-5-3-12(23-24(19,21)22)10-11(13)2-4-15(14)16(18)6-7-17(18)20/h3,5,10,14-16H,2,4,6-9H2,1H3,(H2,19,21,22)/t14-,15-,16+,18+/m1/s1

InChI key

RVKFQAJIXCZXQY-CBZIJGRNSA-N

Application

Lead compound for design of steroid sulfatase inhibitors.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Active site directed inhibition of estrone sulfatase by nonsteroidal coumarin sulfamates.
L W Woo et al.
Journal of medicinal chemistry, 39(7), 1349-1351 (1996-03-29)
A Purohit et al.
Biochemistry, 34(36), 11508-11514 (1995-09-12)
Steroid sulfatases are responsible for the hydrolysis of 3beta-hydroxy steroid sulfates, such as cholesterol and pregnenolone sulfate, and have an important role in regulating the synthesis of estrogenic steroids, from estrone sulfate and dehydroepiandrosterone sulfate, in endocrine-dependent tumors. Although little

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service