Skip to Content
Merck
All Photos(1)

Key Documents

E9402

Sigma-Aldrich

EphB2/Fc Chimera from mouse

>90% (SDS-PAGE), recombinant, expressed in NSO cells, lyophilized powder

Synonym(s):

Cek5, Drt, ERK, Hek5, Nuk, Qek2, Sek3, Tyro5

Sign Into View Organizational & Contract Pricing

Select a Size

200 μG
$842.00

$842.00


Estimated to ship on06 May 2025


Request a Bulk Order

Select a Size

Change View
200 μG
$842.00

About This Item

MDL number:
UNSPSC Code:
51111800
NACRES:
NA.32

$842.00


Estimated to ship on06 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

recombinant

expressed in NSO cells

Assay

>90% (SDS-PAGE)

form

lyophilized powder

potency

0.04-2.5 ng/mL

mol wt

monomer calculated mol wt 85.3 kDa
100-110 kDa by SDS-PAGE (reducing)

packaging

pkg of 200 μg

storage condition

avoid repeated freeze/thaw cycles

technique(s)

ligand binding assay: suitable

impurities

endotoxin, tested

UniProt accession no.

storage temp.

−20°C

Gene Information

mouse ... Ephb2(13844)

General description

EphB2 (Epherin receptor B2) is a receptor tyrosine kinase (RTK), which belongs to a 13 member family of ephrin receptors. This family is divided into two classes A and B. EphB2 belongs to EphB group, which contains five members. The ligands to this group of receptors are named ephrin-B1-B3. It is a bidirectional functioning membrane protein.[1]

Application

EphB2/Fc Chimera from mouse has been used for the investigation of the role of ephrinB-EphB signaling in the induction and maintenance of hyperalgesia, hyperexcitability of neurons of DRG (dorsal root ganglion) and hyperexcitability and elevated synaptic plasticity of neurons of DH (dorsal horn) regions of brain.[1]

Biochem/physiol Actions

EphB2 (Epherin receptor B2) is involved in the development and plasticity of glutamatergic synapses, and their interaction with NMDA receptor. It can also regulate the processing of acute inflammatory pain, and mechanical allodynia as a result of nerve crush. Activation of this receptor results in recruitment and activation of Src-protein kinases. These, in turn, phosphorylate and activate NMDA receptor.[1] It regulates the function of platelets and the development of thrombus. It is also responsible for the retraction of clots, and modulates independent platelet function in a contact-independent manner.[2] EphB2 signaling is also involved in the modulation of axonal sprouting, which is induced by lesions.[3]
Member of the Eph receptor tyrosine kinase family shown to bind ephrin-B1, ephrin-B2, and ephrin-B3; involved in pattern formation and morphogenesis, and in arterial/venous demarcation of the developing vasculature.

Other Notes

Extracellular domain of mouse EphB2 (amino acids 1-548) fused to the C-terminal 6X histidine-tagged Fc region of human IgG1 via a polypeptide linker.

Physical form

Lyophilized from a 0.2 μm filtered solution in 20 mM Tris, pH 8.0.

Analysis Note

The biological activity is measured by its ability to bind recombinant mouse ephrin-B2/Fc in an ELISA assay.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sakthivel Vaiyapuri et al.
Blood, 125(4), 720-730 (2014-11-06)
The Eph kinases, EphA4 and EphB1, and their ligand, ephrinB1, have been previously reported to be present in platelets where they contribute to thrombus stability. Although thrombus formation allows for Eph-ephrin engagement and bidirectional signaling, the importance specifically of Eph
Wen-Tao Liu et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 23(1), 90-98 (2008-09-06)
Cellular and molecular mechanisms underlying opioid tolerance and dependence remain elusive. We investigated roles of EphB receptor tyrosine kinases--which play important roles in synaptic connection and plasticity during development and in the matured nervous system--in development and maintenance of physical
Paul A Nakamura et al.
Neural development, 8, 2-2 (2013-02-06)
Studies of developmental plasticity may provide insight into plasticity during adulthood, when neural circuitry is less responsive to losses or changes in input. In the mammalian auditory brainstem, globular bushy cell axons of the ventral cochlear nucleus (VCN) innervate the
Xue-Jun Song et al.
Pain, 139(1), 168-180 (2008-05-02)
Bidirectional signaling between ephrins and Eph receptor tyrosine kinases was first found to play important roles during development, but recently has been implicated in synaptic plasticity and pain processing in the matured nervous system. We show that ephrinB-EphB receptor signaling
Yuan Han et al.
Molecular pain, 4, 60-60 (2008-11-26)
EphB receptor tyrosine kinases, which play important roles in synaptic connection and plasticity during development and in matured nervous system, have recently been implicated in processing of pain after nerve injury and morphine dependence. Subtypes of the EphB receptors that

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service