Skip to Content
Merck
All Photos(1)

Key Documents

E9252

Sigma-Aldrich

β-Estradiol 17-hemisuccinate

Synonym(s):

1,3,5(10)-Estratriene-3,17β-diol 17-hemisuccinate, 3,17β-Hydroxy-1,3,5(10)-estratriene 17-hemisuccinate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H28O5
CAS Number:
Molecular Weight:
372.45
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
form:
powder
Assay:
≥98.00% (TLC)

Assay

≥98.00% (TLC)

form

powder

solubility

methanol: 9.80-10.20 mg/mL, clear, colorless to faintly yellow

shipped in

ambient

storage temp.

room temp

SMILES string

[H][C@]12CC[C@]3(C)[C@H](CC[C@@]3([H])[C@]1([H])CCc4cc(O)ccc24)OC(=O)CCC(O)=O

InChI

1S/C22H28O5/c1-22-11-10-16-15-5-3-14(23)12-13(15)2-4-17(16)18(22)6-7-19(22)27-21(26)9-8-20(24)25/h3,5,12,16-19,23H,2,4,6-11H2,1H3,(H,24,25)/t16-,17-,18+,19+,22+/m1/s1

InChI key

YJPIDPAGJSWWBE-FNIAAEIWSA-N

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K W Kan et al.
Calcified tissue international, 35(6), 740-744 (1983-09-01)
Using the dextran charcoal method, a specific steroid receptor for dexamethasone, but not 17 beta-estradiol or 17 alpha-methyltrienolone, was demonstrated in cytosol prepared from mid-gestation fetal calf growth cartilage. Zonal analysis of cytoplasmic receptor levels showed that tissue sections from
S S Smith et al.
Brain research, 422(1), 40-51 (1987-09-29)
The purpose of this study was to test whether 17 beta-estradiol (E2) could alter neuronal activity or responsiveness to iontophoretically applied amino acid neurotransmitters in an area not reported to contain classical E2 receptors. Such a region is the cerebellum
C Christiansen et al.
Calcified tissue international, 35(6), 720-722 (1983-09-01)
Bone loss in postmenopausal women can be treated by hormonal substitution therapy. We investigated whether rapid bone losers (as opposed to nonrapid losers) respond sufficiently to estrogen treatment. A sample of 88 early postmenopausal women was followed up for a
K Kondo et al.
Endocrinologia japonica, 27(3), 307-313 (1980-06-01)
Effects of estradiol hemisuccinate and diethylstilbestrol diphosphate on the cardiovascular system in male rats were examined in vivo and in vitro. In conscious rats, intravenously administered estradiol or diethylstilbestrol inhibited vascular responses to norepinephrine in a dose-dependent manner. Diethylstilbestrol in
T L Thompson et al.
Journal of neurochemistry, 62(5), 1750-1756 (1994-05-01)
The ability of estrogen to modulate mesolimbic dopamine (DA) was examined using in vivo voltammetry. Estrogen priming (5 micrograms, 48 h) of ovariectomized (ovx) female rats resulted in a slight decrease in K(+)-stimulated DA release measured in the nucleus accumbens:

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service