Skip to Content
Merck
All Photos(2)

Key Documents

E8283

Sigma-Aldrich

pFLAG-CTS Expression Vector

Bacterial vector for periplasmic expression of C-terminal FLAG fusion proteins

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

tag

FLAG® tagged

grade

Molecular Biology
for molecular biology

form

buffered aqueous solution

shipped in

dry ice

storage temp.

−20°C

General description

The pFLAG-CTS Expression Vector is a 5.3 kb E. coli expression vector used for cytoplasmic expression of a properly inserted open reading frame as a C-terminal FLAG® fusion protein. The FLAG epitope is a small, hydrophilic 8 amino acid tag (DYKDDDDK) that provides for sensitive detection and high quality purification using ANTI-FLAG products.

C-terminal FLAG fusion proteins may be purified using Monoclonal ANTI-FLAG M2, Catalog Number F3165, and ANTI-FLAG M2 Affinity Gel, Catalog Number A2220.

The pFLAG-CTS-BAP Control Plasmid is a 6.7 kb E. coli plasmid used for efficient and controlled periplasmic expression of C-terminal FLAG-BAP fusion protein.

Vector Maps and Sequences

Application

The pFLAG-CTS Expression Vector is suitable for cloning and expression of C-terminal FLAG® fusion proteins in E. coli.

Components

  • pFLAG-CTS Expression Vector 10 μg (E5269) is supplied as 0.5 mg/ml in 10 mM Tris-HCl (pH 8.0) with 1 mM EDTA.
  • pFLAG-CTS-BAP Control Plasmid 1 μg (P7707) is supplied as 0.5 mg/ml in 10 mM Tris-HCl (pH 8.0) with 1 mM EDTA.

Principle

The promoter-regulatory region of the strong tac promoter (a hybrid of the trp and lac promoters from E.coli) drives transcription of ORF-FLAG fusion constructs. Control of transcription is regulated by the presence of the lacO sequences and inclusion of the lac repressor gene (lacI) on the plasmid.

Legal Information

FLAG is a registered trademark of Merck KGaA, Darmstadt, Germany
pFLAG-CTS is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G A Smith et al.
Molecular microbiology, 17(5), 945-951 (1995-09-01)
Listeria monocytogenes is a Gram-positive facultative intracytoplasmic bacterial pathogen that exhibits rapid actin-based motility in eukaryotic cells and in cell-free cytoplasmic extracts. The protein product of the actA gene is required for bacterial movement and is normally expressed in a
Saadet Albayrak Guralp et al.
PloS one, 8(3), e59305-e59305 (2013-03-26)
Antimicrobial peptides (AMPs) belong to a class of natural microbicidal molecules that have been receiving great attention for their lower propensity for inducing drug resistance, hence, their potential as alternative drugs to conventional antibiotics. By generating AMP libraries, one can
H A de Boer et al.
Proceedings of the National Academy of Sciences of the United States of America, 80(1), 21-25 (1983-01-01)
Two hybrid promoters that are functional in Escherichia coli have been constructed. These hybrid promoters, tacI and tacII, were derived from sequences of the trp and the lac UV5 promoters. In the first hybrid promoter (tacI), the DNA upstream of
Montarop Yamabhai et al.
Journal of biotechnology, 133(1), 50-57 (2007-10-24)
Bacillus spp. are Gram-positive bacteria that secrete a large number of extracellular proteins of industrial relevance. In this report, three Bacillus extracellular hydrolytic enzymes, i.e., alpha-amylase, mannanase and chitinase, were cloned and over-expressed in Gram-negative Escherichia coli. We found that
Parisa Asvadi et al.
Journal of molecular recognition : JMR, 15(5), 321-330 (2002-11-26)
In an attempt to generate recombinant anti-D reagents for possible diagnostic and therapeutic use we cloned the genes encoding the variable (V) domains of a human anti-D antibody secreted by the lymphoblastoid cell line BTSN4. A single-chain Fv (scFv) fragment

Related Content

Bacterial Expression Vectors: tac Promoter System

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service