E8022
pRc/RSV Expression vector
set of expression vectors powered by either the CMV2 or the RSV promoter
About This Item
Recommended Products
grade
Molecular Biology
for molecular biology
form
buffered aqueous solution
storage temp.
−20°C
Related Categories
General description
- The polyadenylation signal and transcription termination sequence from BGH to enhance mRNA stability
- The SV40 replication origin for episomal replication
- The f1 origin for rescue of the sense strand
- The neomycin resistance gene, and
- An ampicillin resistance gene and colE1 origin for selection and maintenance in E. coli.
The primary difference between these expression vectors is that pRc contains the enhancer-promoter sequence from human cytomegalovirus (CMV2) and pRSV from Rous sarcoma virus (RSV). Both are strong promoters for high-level transcription.
The pRc MCS contains unique ApaI, HindIII, NotI, and XbaI restriction sites, plus redundant BstXI sites.
The pRSV MCS contains unique HindIII, NotI, SpeI, and XbaI restriction sites, plus redundant BstXI sites.
Specificity
Features and Benefits
- the polyadenylation signal and transcription termination sequence from BGH to enhance mRNA stability
- the SV40 replication origin for episomal replication
- the f1 origin for rescue of the sense strand
- the neomycin resistance gene, and
- an ampicillin resistance gene and colE1 origin for selection and maintenance in E. coli.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service