E7642
Endoglycosidase H from Streptomyces plicatus
recombinant, expressed in E. coli, buffered aqueous solution
Synonym(s):
β-N-Acetylglucosaminidase H
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
CAS Number:
MDL number:
UNSPSC Code:
12352204
recombinant
expressed in E. coli
form
buffered aqueous solution
mol wt
27 kDa
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
Unit Definition
One unit will hydrolyze 1.0 μmole of dabsyl-Asn-(GlcNAc)2(Man)5 per min at pH 5.5 at 37 °C.
Physical form
Solution in 0.05 M sodium phosphate, pH 7, containing 25 mM EDTA and preservative
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Rhizomucor miehei aspartic proteinases having improved properties.
M K Harboe
Advances in experimental medicine and biology, 436, 293-296 (1998-04-30)
M V Padkina et al.
Prikladnaia biokhimiia i mikrobiologiia, 46(4), 448-455 (2010-09-29)
The HuIFNA16, HuIFNB, and BoIFNG genes encoding human [alpha]16, beta-interferons and bovine gamma-interferon were cloned under the control of the yeast Pichia pastoris AOX1 gene promoter. The yeast strains producing heterologous interferons intracellularly and extracellularly were constructed. There was no
Valentina Botti et al.
PloS one, 6(8), e23838-e23838 (2011-08-23)
Hepatitis C Virus E1E2 heterodimers are components of the viral spike. Although there is a general agreement on the necessity of the co-expression of both E1 and E2 on a single coding unit for their productive folding and assembly, in
Yusuke Tomabechi et al.
Carbohydrate research, 345(17), 2458-2463 (2010-10-12)
To determine the structural specificity of the glycosyl acceptor of the transglycosylation reaction using endo-β-N-acetylglucosaminidase (ENGase) (EC 3.2.1.96) from Mucor hiemalis (Endo-M), several acceptor derivatives were designed and synthesized. The narrow regions of the 1,3-diol structure from the 4- to
Roger S Zou et al.
Aging, 3(10), 968-984 (2011-10-13)
A distinct conformational transition from the α-helix-rich cellular prion protein (PrPC) into its β-sheet-rich pathological isoform (PrPSc) is the hallmark of prion diseases, a group of fatal transmissible encephalopathies that includes spontaneous and acquired forms. Recently, a PrPSc-like intermediate form
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service