Skip to Content
Merck
All Photos(1)

Key Documents

E7153

Sigma-Aldrich

Anti-phospho-ERK5 (pThr218/pTyr220) antibody produced in rabbit

affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-BMK1

Sign Into View Organizational & Contract Pricing

Select a Size

1 VIAL
$1,030.00

$1,030.00


Estimated to ship on31 May 2025



Select a Size

Change View
1 VIAL
$1,030.00

About This Item

EC Number:
MDL number:
UNSPSC Code:
12352203
NACRES:
NA.44

$1,030.00


Estimated to ship on31 May 2025


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

species reactivity

human

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... MAPK7(5598)

General description

Extracellular Signal-Regulated Kinases (ERKs) are proteins that belong to mitogen-activated protein kinase superfamily (MAPK) and have a pivotal role in cell cycle, differentiation, transcription and learning and memory through signal transduction in the cytoplasm and the nucleus. ERK5 MAP kinase cascade has a crucial functions like- induction of special form of cardiac hypertrophy, enhanced neuronal survival and early steps of muscle cell differentiation. Anti-phospho-ERK5 (pThr218/pTyr220) antibody can be used in immunostaining. Rabbit anti- phospho-ERK5 (pThr218/pTyr220) antibody reacts specifically with ERK5 (88 kD) of human.

Immunogen

Synthetic phosphopeptide derived from the region of human ERK5 that contains threonine 218 and tyrosine 220. This sequence is conserved in human and mouse.

Application

Each vial contains enough material for 10 blots. The end user should determine the correct working dilution for their process.

Physical form

Solution in Dulbecco′s PBS, pH 7.3, with 50% glycerol, 1 mg/ml BSA and ≤ 0.1% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Congde Chen et al.
Aging, 11(2), 697-706 (2019-01-27)
Diabetic patients are prone to developing Alzheimer's disease (AD), in which microglia play a critical role. However, the direct effect of high glucose (HG) on microglia and the role of extracellular-signal-regulated kinase 5 (ERK5) signaling in this interaction have not
Kaoru Toshima et al.
International immunology, 31(4), 211-223 (2018-12-19)
Sphingomyelin (SM) in combination with cholesterol forms specialized membrane lipid microdomains in which specific receptors and signaling molecules are localized or recruited to mediate intracellular signaling. SM-microdomain levels in mouse thymus were low in the early CD4+CD8+ double-positive (DP) stage
Suji Kim et al.
Scientific reports, 8(1), 15172-15172 (2018-10-13)
Recent studies have indicated that protease-activated receptor-1 (PAR-1) is involved in cytoprotective and anti-inflammatory responses in endothelial cells (ECs). However, the role of PAR-1 in laminar flow-mediated atheroprotective responses remains unknown. Herein, we investigated whether PAR-1 regulates laminar flow-mediated mechanotransduction
Mahmud Hasan et al.
Molecular pharmacology, 96(2), 272-296 (2019-06-22)
Tamoxifen is used to prevent and treat estrogen receptor-positive (ER+) breast cancer (BC); however, its chronic use can increase uterine cancer risk and induce tamoxifen resistance. Novel melatonin-tamoxifen drug conjugates may be promising to treat BC and may help offset
Mingming Zhu et al.
Environmental toxicology and pharmacology, 63, 29-33 (2018-08-21)
Prostate cancer is one of the most commonly diagnosed cancers in man. Studies have shown that phthalates may act as promoters in various types of cancer; however, the role of phthalates in prostate cancer has been rarely reported. The MAPK/AP-1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service