Skip to Content
Merck
All Photos(3)

Key Documents

E7138

Sigma-Aldrich

Ethosuximide

Synonym(s):

2-Ethyl-2-methylsuccinimide

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$95.70
100 G
$656.00

$95.70


Estimated to ship on18 March 2025

New, lower price on this item!

Request a Bulk Order

Select a Size

Change View
5 G
$95.70
100 G
$656.00

About This Item

Empirical Formula (Hill Notation):
C7H11NO2
CAS Number:
Molecular Weight:
141.17
EC Number:
MDL number:
UNSPSC Code:
12352111
PubChem Substance ID:
NACRES:
NA.77

$95.70


Estimated to ship on18 March 2025

New, lower price on this item!

Request a Bulk Order

Quality Level

solubility

ethanol: 100 mg/mL

SMILES string

O=C1NC(CC1(C)CC)=O

InChI

1S/C7H11NO2/c1-3-7(2)4-5(9)8-6(7)10/h3-4H2,1-2H3,(H,8,9,10)

InChI key

HAPOVYFOVVWLRS-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

Ethosuximide has been used:
  • to confirm its effect on spike wave discharges (SWD) in rats[1]
  • to examine its effects on spontaneous locomotion and anxiety-related thigmotaxis behaviours in zebrafish larvae of wild type[2]
  • as a model drug for the treatment of epilepsy in vivo(4)

Biochem/physiol Actions

Ethosuximide is an anticonvulsant drug[3] and an antagonist for T-type calcium channel. It is known to prevent spike wave discharges, characterized in absence seizures.[1]

Features and Benefits

This compound is featured on the Calcium Channels page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Self-assembling PVA-F127 thermosensitive nanocarriers with highly sensitive magnetically-triggered drug release for epilepsy therapy in vivo
Huang HY, et al.
Journal of Materials Chemistry, 22(17), 8566-8573 (2012)
Emilio Russo et al.
Epilepsia, 52(7), 1341-1350 (2011-06-04)
Depression is most commonly associated with epilepsy. Recent reports have suggested a putative relationship between seizure development and onset of depressive behavior, whereas others proposed that two clinical entities might represent different neuropathologic aspects of the same neurologic disorder. The
Gabi Dezsi et al.
Epilepsia, 54(4), 635-643 (2013-03-08)
Ethosuximide (ESX) is a drug of choice for the symptomatic treatment of absence seizures. Chronic treatment with ESX has been reported to have disease-modifying antiepileptogenic activity in the WAG/Rij rat model of genetic generalized epilepsy (GGE) with absence seizures. Here
Gilles van Luijtelaar et al.
International journal of psychophysiology : official journal of the International Organization of Psychophysiology, 85(1), 49-54 (2011-09-29)
Recently it was established that early long lasting treatment with the anti-absence drug ethosuximide (ETX) delays the occurrence of absences and reduces depressive-like symptoms in a genetic model for absence epilepsy, rats of the WAG/Rij strain. Here it is investigated
Spike-wave discharges in adult Sprague-Dawley rats and their implications for animal models of temporal lobe epilepsy
Pearce PS, et al.
Epilepsy & Behavior : E&B, 32, 121-131 (2014)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service