Skip to Content
Merck
All Photos(1)

Key Documents

E7134

Sigma-Aldrich

Evening primrose oil

Synonym(s):

Oenothera biennis

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352211

refractive index

n20/D 1.479

density

0.926 g/mL at 25 °C

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

A crude source of γ-linolenic acid (5-15%). Also contains linoleic, oleic, palmitic and stearic acids.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service