E6886
5-Ethyluracil
Synonym(s):
2,4-Dihydroxy-5-ethylpyrimidine, Homothymine
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
mp
>300 °C (lit.)
SMILES string
CCC1=CNC(=O)NC1=O
Looking for similar products? Visit Product Comparison Guide
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Biological chemistry, 378(5), 407-415 (1997-05-01)
Steady-state kinetics of the N6-adenine Dam methyltransferase have been measured using as substrates non-self-complementary tetradecanucleotide duplexes that contain the GATC target sequence. Modifications in the GATC target sequence of one or both of the strands included substitution of guanine by
Nucleic acids research, 16(11), 4761-4776 (1988-06-10)
Decadeoxyribonucleotides containing uracil, 5-bromouracil, 5-cyanouracil and 5-ethyluracil in recognition sequences of restriction endonucleases Bgl II, Sau 3AI, Mbo I were synthesized. Decanucleotides containing 5-bromouracil in place of thymine had essentially the same susceptibility to all the restriction endonucleases. Uracil-containing decanucleotides
Molecular diversity, 15(3), 677-686 (2010-11-17)
Oligoribonucleotides carrying 5-ethyluridine units were prepared using solid-phase phosphoramidite chemistry. The introduction of the tert-butyldimethylsilyl group at the 2'-OH position proceeded in good yield and very high 2'-regioselectivity. RNA duplexes carrying 5-ethyluridine either at the sense or the guide strands
Nucleic acids research, 23(18), 3648-3655 (1995-09-25)
We have measured steady-state kinetics of the N6-adenine methyltransferase Dam Mtase using as substrates non-selfcomplementary tetradecamer duplexs (d[GCCGGATCTAGACG]-d[CGTCTAGATCC-GGC]) containing the hemimethylated GATC target sequence in one or the other strand and modifications in the GATC target sequence of the complementary
Clinical cancer research : an official journal of the American Association for Cancer Research, 1(9), 991-996 (1995-09-01)
Dihydropyrimidine dehydrogenase (DPD) is the rate-limiting enzyme of 5-fluorouracil (FU) catabolism. Ethynyluracil (776C) is a very potent, mechanism-based irreversible DPD inhibitor that improves the antitumor efficacy and the therapeutic index of FU in laboratory animals. We tested the cytotoxic effects
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service