Skip to Content
Merck
All Photos(1)

Key Documents

E6878

Sigma-Aldrich

Endoglycosidase H from Streptomyces griseus

lyophilized powder

Synonym(s):

β-N-Acetylglucosaminidase H

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204

form

lyophilized powder

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Unit Definition

One unit will hydrolyze 1.0 μmole of N-acetyl-(14C)Asn(GlcNAc)2(Man)5 per min at pH 5.0 at 37 °C.

Physical form

Lyophilized powder containing bovine serum albumin

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liv Anette Bøhle et al.
FEMS microbiology letters, 325(2), 123-129 (2011-11-19)
It has been demonstrated previously that Enterococcus faecalis produces secreted endoglycosidases that enable the bacteria to remove N-linked glycans from glycoproteins. One enzyme potentially responsible for this activity is EF0114, comprising a typical GH18 endoglycosidase domain and a GH20 domain.
Helena Ryšlavá et al.
The FEBS journal, 278(14), 2469-2484 (2011-05-14)
Fungal β-N-acetylhexosaminidases are inducible extracellular enzymes with many biotechnological applications. The enzyme from Penicillium oxalicum has unique enzymatic properties despite its close evolutionary relationship with other fungal hexosaminidases. It has high GalNAcase activity, tolerates substrates with the modified N-acyl group
Wei Zhang et al.
Talanta, 85(1), 499-505 (2011-06-08)
Endoglycosidase is a class of glycosidases that specifically cleaves the glycosidic bond between two proximal residues of GlcNAc in the pentasaccharide core of N-glycan, leaving the innermost GlcNAc still attached to its parent protein, which provides a different diagnostic maker
John E Schiel et al.
Journal of mass spectrometry : JMS, 46(7), 649-657 (2011-06-28)
The current project describes the chemoenzymatic modification of bovine ribonuclease B (RNase B) to contain a single glycosylation site with a known glycan. A reactive disaccharide oxazoline derivative was synthesized and stereospecifically added to deglycosylated RNase B through endo-β-N-acetylglucosaminidase M
Shu-Quan Fan et al.
The Journal of biological chemistry, 287(14), 11272-11281 (2012-02-10)
Endo-β-N-acetylglucosaminidase from Streptococcus pneumoniae (Endo-D) is an endoglycosidase capable of hydrolyzing the Fc N-glycan of intact IgG antibodies after sequential removal of the sialic acid, galactose, and internal GlcNAc residues in the N-glycan. Endo-D also possesses transglycosylation activity with sugar

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service