Skip to Content
Merck
All Photos(1)

Key Documents

E6627

Sigma-Aldrich

cis-5,8,11,14,17-Eicosapentaenoic acid sodium salt

≥99% (capillary GC)

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$85.00
25 MG
$229.00

$85.00


Available to ship on31 March 2025Details


Request a Bulk Order

Select a Size

Change View
5 MG
$85.00
25 MG
$229.00

About This Item

Linear Formula:
CH3(CH2CH=CH)5(CH2)3CO2Na
CAS Number:
Molecular Weight:
324.43
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.77

$85.00


Available to ship on31 March 2025Details


Request a Bulk Order

biological source

synthetic (organic)

Quality Level

Assay

≥99% (capillary GC)

form

film or powder

color

white to off-white

solubility

methanol: 50 mg/mL, clear to slightly hazy, colorless to faintly yellow

shipped in

dry ice

storage temp.

−20°C

SMILES string

[Na].CC\C=C/C\C=C/C\C=C/C\C=C/C\C=C/CCCC(O)=O

InChI

1S/C20H30O2.Na.H/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20(21)22;;/h3-4,6-7,9-10,12-13,15-16H,2,5,8,11,14,17-19H2,1H3,(H,21,22);;/b4-3-,7-6-,10-9-,13-12-,16-15-;;

InChI key

TVEGTERCKGGVCJ-LIJGWLRISA-N

Gene Information

human ... ALOX5(240)
mouse ... ALOX5(11689)
rat ... ALOX5(25290)

Looking for similar products? Visit Product Comparison Guide

General description

Eicosapentaenoic acid (EPA)/timnodonic acid is an omega-3 polyunsaturated fatty acid (PUFA). This long-chain fatty acid is mainly found in fish oil.

Application

cis-5,8,11,14,17-Eicosapentaenoic acid sodium salt has been used:
  • to study its effect on the pathology of lupus in drug-induced and spontaneous mouse models
  • to pretreat mouse alveolar macrophages to study the effect/relationship between amiodarone and eicosapentaenoic acid (EPA) concerning the cytokine release
  • to examine if pharmacological correction with EPA supplementation might restore autophagic flux and reduce renal lipotoxicity in mice

Biochem/physiol Actions

5-Lipoxygenase inhibitor; reduces thromboxane A2 production.
Eicosapentaenoic acid (EPA) possesses anti-hyperglycemic, anti-oxidant, anti-hyperlipidemic, and anti-inflammatory properties. It aids in improving vasoreactivity. EPA is capable of suppressing vascular smooth muscle proliferation. It ameliorates pulmonary hypertension by preventing Fyn kinase activity. EPA exhibits a beneficial effect on lipid metabolism. It is used to treat hypercholesterolemia and atherosclerosis.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lin Hai Kurahara et al.
Journal of molecular and cellular cardiology, 148, 50-62 (2020-09-06)
Pulmonary arterial hypertension (PAH) is a multifactorial disease characterized by pulmonary arterial vasoconstriction and remodeling. Src family tyrosine kinases, including Fyn, play critical roles in vascular remodeling via the inhibition of STAT3 signaling. EPA is known to inhibit Fyn kinase
Y Futamura
Japanese journal of pharmacology, 69(4), 335-341 (1995-12-01)
I studied the effect of amiodarone, a cationic amphiphilic drug, on the cytokine release and protein kinase C (PKC) activity of mouse alveolar macrophages. In addition, I examined the relationship between amiodarone and eicosapentaenoic acid (EPA) with respect to the
Azusa Kobayashi et al.
Frontiers in immunology, 12, 650856-650856 (2021-07-03)
Accumulating evidence suggests that cholesterol accumulation in leukocytes is causally associated with the development of autoimmune diseases. However, the mechanism by which fatty acid composition influences autoimmune responses remains unclear. To determine whether the fatty acid composition of diet modulates
Yong-Ping Zhang et al.
Prostaglandins, leukotrienes, and essential fatty acids, 136, 85-94 (2017-08-06)
Omega-3 polyunsaturated fatty acids (n-3 PUFAs), docosahexaenoic acid (DHA) and eicosapentaenoic acid (EPA), have been reported to prevent neurodegenerative diseases such as Alzheimer's disease (AD) in both experimental and clinical/epidemiological studies. However, whether DHA and EPA from natural products exert
Takeshi Yamamoto et al.
Autophagy, 17(7), 1700-1713 (2020-06-18)
Recently, we identified a novel mechanism of lipotoxicity in the kidney proximal tubular cells (PTECs); lipid overload stimulates macroautophagy/autophagy for the renovation of plasma and organelle membranes to maintain the integrity of the PTECs. However, this autophagic activation places a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service