Skip to Content
Merck
All Photos(1)

Key Documents

E5376

Supelco

Etiocholan-17β-ol-3-one

Synonym(s):

17β-Hydroxy-5β-androstan-3-one, 17β-Hydroxyetiocholan-3-one, 5β-Androstan-17β-ol-3-one, 5β-Dihydrotestosterone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H30O2
CAS Number:
Molecular Weight:
290.44
MDL number:
UNSPSC Code:
41116107

drug control

Home Office Schedule 4.2; regulated under CDSA - not available from Sigma-Aldrich Canada

SMILES string

CC12CCC3C(CCC4CC(=O)CCC34C)C1CCC2O

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 1B - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service