Skip to Content
Merck
All Photos(7)

Key Documents

E4913

Sigma-Aldrich

Plant/Fungi RNA Purification Kit

greener alternative

sufficient for 50 purifications

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105501
NACRES:
NA.52

usage

sufficient for 50 purifications

greener alternative product characteristics

Designing Safer Chemicals
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

greener alternative category

General description

Norgen Biotek′s Plant/Fungi Total RNA Purification Kit provides a rapid method for the isolation and purification of total RNA, including viral RNA, from a wide range of plant and filamentous fungal species. Total RNA can be purified from fresh or frozen plant tissues, plant cells or filamentous fungi samples using this kit. All sizes of RNA are purified, from large mRNA and ribosomal RNA down to microRNA (miRNA) and small interfering RNA (siRNA). The procedure is rapid and convenient, as it does not rely on the use of liquid nitrogen in order to homogenize the samples.

Application

For the rapid purification of total RNA (including microRNA) from plants and fungi.
Purified RNA is of the highest quality and can be used in a number of downstream applications, including:
  • qRT-PCR
  • Reverse transcription PCR
  • Northern blotting
  • RNAse protection
  • Primer extension
  • Expression array assays
  • Next Generation Sequencing

Features and Benefits

  • Isolate RNA from a wide range of plant and filamentous fungi samples.
  • Robust lysis buffer is suitable for challenging samples such as pine needle or grape leaf.
  • Isolate a diversity of RNA species from large mRNA, microRNA, and viral RNA.
  • Purification of high yield and quality RNA.
  • No phenol/chloroform extraction required.
  • Liquid nitrogen is not required for homogenization

Other Notes

Storage Conditions and Product Stability

All solutions should be kept tightly sealed and stored at room temperature. These reagents should remain stable for at least 1 year in their unopened containers.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has Inherently Safer Chemistry, compared to the standard use of phenol and chloroform to perform DNA extractions.

related product

Product No.
Description
Pricing

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jocelyn P S Chua et al.
The New phytologist, 196(3), 862-872 (2012-08-31)
Development of the symbiotic association in the bipartite lichen Pseudocyphellaria crocata was investigated by characterizing two regions of the thallus. Thallus organization was examined using microscopy. A HIP1-based differential display technique was modified for use on Nostoc strains, including lichenized
Michaela Novodvorska et al.
BMC genomics, 14, 246-246 (2013-04-13)
Genome-wide analysis was performed to assess the transcriptional landscape of germinating A. niger conidia using both next generation RNA-sequencing and GeneChips. The metabolism of storage compounds during conidial germination was also examined and compared to the transcript levels from associated
Moreno Colaiacovo et al.
Frontiers in plant science, 3, 165-165 (2012-08-03)
MicroRNAs (miRNAs) are short non-coding RNA molecules produced from hairpin structures and involved in gene expression regulation with major roles in plant development and stress response. Although each annotated miRNA in miRBase (www.mirbase.org) is a single defined sequence with no
Yuan Song et al.
Biochemical and biophysical research communications, 396(2), 187-192 (2010-03-25)
DNA methyltransferase2 (DNMT2) is always deemed to be enigmatic, because it contains highly conserved DNA methyltransferase motifs but lacks the DNA methylation catalytic capability. Here we show that Arabidopsis DNA methyltransferase2 (AtDNMT2) is localized in nucleus and associates with histone
I El-Sharkawy et al.
Journal of experimental botany, 63(3), 1225-1239 (2011-11-15)
Several dwarf plum genotypes (Prunus salicina L.), due to deficiency of unknown gibberellin (GA) signalling, were identified. A cDNA encoding GA 2-oxidase (PslGA2ox), the major gibberellin catabolic enzyme in plants, was cloned and used to screen the GA-deficient hybrids. This

Protocols

The Plant RNA/DNA Purification Kit provides a rapid method for the isolation and purification of total RNA and DNA simultaneously from a single sample of plants.

The Plant/Fungi DNA Isolation Kit provides a rapid method for the isolation and purification of total DNA from a wide range of plant and fungi species.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service