Skip to Content
Merck
All Photos(1)

Key Documents

E3526

Sigma-Aldrich

Anti-phospho-Elk-1 (pSer383) antibody produced in rabbit

affinity isolated antibody, buffered aqueous glycerol solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

species reactivity

rat, mouse, human

technique(s)

western blot (chemiluminescent): 1:1,000 using Elk-1 fusion protein phosphorylated with purified Erk2

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pSer383)

Gene Information

human ... ELK1(2002)
mouse ... Elk1(13712)
rat ... Elk1(314436)

Specificity

The antibody detects Elk-1 protein only when phosphorylated at Ser383.

Immunogen

synthetic phosphopeptide corresponding to residues around phosphorylated Ser383 of human Elk-1.

Application

Anti-phospho-Elk-1 (pSer383) antibody produced in rabbit is suitable for western blotting (chemiluminescent) at a dilution of 1:1,000 using Elk-1 fusion protein phosphorylated with purified Erk2.

Biochem/physiol Actions

ETS domain-containing protein Elk-1 is a protein encoded by the ELK1 gene in humans. It is a member of the Ets family of transcription factors. Elk-1 activation is important for executing its physiologic functions. Its activation is positively associated with ER and with Cyclin D1. Elk1 plays a novel role in controlling cell migration. Elk-1 is involved in a number of important cell processes through the activation of different genes in a number of normal tissues as well as in many malignancies. It is also plays a role in breast neoplasia.

Physical form

Solution in 10 mM sodium HEPES, pH 7.5, containing 150 mM sodium chloride, 100 μg/ml bovine serum albumin and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rachasak Boonhok et al.
Proceedings of the National Academy of Sciences of the United States of America, 113(25), E3519-E3528 (2016-05-18)
IFN-γ is a major regulator of immune functions and has been shown to induce liver-stage Plasmodium elimination both in vitro and in vivo. The molecular mechanism responsible for the restriction of liver-stage Plasmodium downstream of IFN-γ remains uncertain, however. Autophagy
Jacqueline F Morris et al.
Human pathology, 44(5), 766-776 (2012-11-02)
Elk-1 is a member of the Ets family of transcription factors, which are identified by a conserved Ets DNA-binding domain that mediates transcriptional regulation at Ets sequence--containing promoters. The activation domain of Elk-1 is important for executing its physiologic functions
Aggelos Laliotis et al.
Breast (Edinburgh, Scotland), 22(1), 89-95 (2012-11-07)
Recently an increased interest on Elk1 protein and its role in breast cancer evolution has been noted. This protein is an element of the Ets family of transcription factors and it has been involved in a number of important cell
Zaneta Odrowaz et al.
PLoS genetics, 8(5), e1002694-e1002694 (2012-05-17)
Eukaryotic transcription factors are grouped into families and, due to their similar DNA binding domains, often have the potential to bind to the same genomic regions. This can lead to redundancy at the level of DNA binding, and mechanisms are

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service