Skip to Content
Merck
All Photos(1)

Key Documents

E2661

Sigma-Aldrich

Estropipate

≥98% (HPLC), powder

Synonym(s):

Estropipate, 3-(Sulfooxy)-estra-1,3,5(10)-trien-17-one : piperazine (1:1), Estrone sulfate piperazine salt, Harmogen, Ogen

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$103.00
25 MG
$412.00

$103.00


Estimated to ship on26 May 2025

New, lower price on this item!

Request a Bulk Order

Select a Size

Change View
5 MG
$103.00
25 MG
$412.00

About This Item

Empirical Formula (Hill Notation):
C18H22O5S·C4H10N2
CAS Number:
Molecular Weight:
436.56
EC Number:
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

$103.00


Estimated to ship on26 May 2025

New, lower price on this item!

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D 100 to 120°, c = 1 in DMSO

color

white to tan

solubility

DMSO: ≥24 mg/mL

originator

Johnson & Johnson

storage temp.

2-8°C

SMILES string

C1CNCCN1.C[C@]23CC[C@H]4[C@@H](CCc5cc(OS(O)(=O)=O)ccc45)[C@@H]2CCC3=O

InChI

1S/C18H22O5S.C4H10N2/c1-18-9-8-14-13-5-3-12(23-24(20,21)22)10-11(13)2-4-15(14)16(18)6-7-17(18)19;1-2-6-4-3-5-1/h3,5,10,14-16H,2,4,6-9H2,1H3,(H,20,21,22);5-6H,1-4H2/t14-,15-,16+,18+;/m1./s1

InChI key

HZEQBCVBILBTEP-ZFINNJDLSA-N

Gene Information

human ... ESR1(2099)

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Estropipate is the piperazine salt of the estrogen receptor agonist estrone-3-sulfate and a potent, specific inhibitor of the organic anion transporting polypeptide 1B1 (OATP1B1, IC50 = 60 nM).
Estropipate is the piperazine salt of the estrogen receptor agonist estrone-3-sulfate and a potent, specific inhibitor of the organic anion transporting polypeptide 1B1.

Features and Benefits

This compound is a featured product for ADME Tox research. Click here to discover more featured ADME Tox products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the Nuclear Receptors (Steroids) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by Johnson & Johnson. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 1A

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Juliana Félix-Silva et al.
PloS one, 9(8), e104952-e104952 (2014-08-16)
Snakebites are a serious public health problem due their high morbi-mortality. The main available specific treatment is the antivenom serum therapy, which has some disadvantages, such as poor neutralization of local effects, risk of immunological reactions, high cost and difficult
Andrea Doni et al.
The Journal of experimental medicine, 212(6), 905-925 (2015-05-13)
Pentraxin 3 (PTX3) is a fluid-phase pattern recognition molecule and a key component of the humoral arm of innate immunity. In four different models of tissue damage in mice, PTX3 deficiency was associated with increased fibrin deposition and persistence, and

Articles

We offers many products related to Nuclear Receptors (Steroids) for your research needs.

Discover Bioactive Small Molecules for ADME/Tox

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service