Skip to Content
Merck
All Photos(1)

Documents

E1408

Sigma-Aldrich

14-3-3 Eta histidine-tagged human

recombinant, expressed in E. coli, C-terminal, ≥90% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

14-3-3η, 14-3-3F, 14-3-3e, MGC93547, Tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, eta polypeptide, YWHAH, Ywha1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

recombinant

expressed in E. coli

Assay

≥90% (SDS-PAGE)

form

buffered aqueous glycerol solution

mol wt

~30 kDa (14-3-3 proteins)

shipped in

dry ice

storage temp.

−20°C

Gene Information

Biochem/physiol Actions

14-3-3 proteins are involved in a multitude of biological processes and play a regulatory role in processes such as apoptotic cell death, mitogenic signal transduction, and cell cycle control. 14-3-3η binds various proteins such as the MTLX-2 transcription factor as well as the DNA binding domain of Miz1 transcription factor. 14-3-3η also binds the glucocorticoid receptor, functioning as a positive regulator in the glucocorticoid signaling pathway. 14-3-3η is expressed and regulated in nerve cells.

Physical form

Supplied as a 0.2 μm filtered solution in 50 mM Tris-HCl, pH 7.6, 1 mM EDTA, 1 mM DTT, and 20% glycerol (w/v).

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S J Tang et al.
The Journal of biological chemistry, 273(39), 25356-25363 (1998-09-17)
Homeodomain proteins play important roles in various developmental processes, and their functions are modulated by polypeptide cofactors. Here we report that both in vitro and in vivo, 14-3-3eta is associated with the TLX-2 homeodomain transcription factor that is required for
Yoon Suk Kim et al.
Endocrinology, 146(7), 3133-3140 (2005-03-26)
The glucocorticoid receptor (GR), a member of the nuclear receptor superfamily, mediates the effects of glucocorticoids. It is known that 14-3-3 family proteins interact with GR and regulate its transcriptional activity. They also bind to several molecules and influence many
José M A Moreira et al.
Molecular & cellular proteomics : MCP, 7(7), 1225-1240 (2008-04-02)
The 14-3-3 proteins constitute a family of highly conserved and broadly expressed multifunctional polypeptides that are involved in a variety of important cellular processes that include cell cycle progression, growth, differentiation, and apoptosis. Although the exact cellular function(s) of 14-3-3
Michael Wanzel et al.
Nature cell biology, 7(1), 30-41 (2004-12-08)
The transcription factor Miz1 is required for DNA-damage-induced cell-cycle arrest. We have now identified 14-3-3eta as a gene that inhibits Miz1 function through interaction with its DNA binding domain. Binding of 14-3-3eta to Miz1 depends on phosphorylation by Akt and
14-3-3 proteins: active cofactors in cellular regulation by serine/threonine phosphorylation.
Guri Tzivion et al.
The Journal of biological chemistry, 277(5), 3061-3064 (2001-11-16)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service