E1032
Anti-EN-RAGE antibody produced in goat
affinity isolated antibody, lyophilized powder
Synonym(s):
Anti-CAGC, Anti-S100 calcium binding protein A12, Anti-S100A12, Anti-p6
Select a Size
$785.00
Estimated to ship on29 May 2025
Select a Size
About This Item
$785.00
Estimated to ship on29 May 2025
Recommended Products
biological source
goat
Quality Level
conjugate
unconjugated
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
form
lyophilized powder
species reactivity
human
technique(s)
flow cytometry: 3-10 μg/mL
immunohistochemistry: 5-15 μg/mL
western blot: 0.1-0.2 μg/mL
UniProt accession no.
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... S100A12(6283)
General description
Immunogen
Application
Biochem/physiol Actions
Physical form
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service