D7821
dPPA
≥98% (HPLC)
Synonym(s):
12-Deoxyphorbol 13-phenylacetate 20-acetate, DOPPA
About This Item
Recommended Products
Quality Level
Assay
≥98% (HPLC)
form
oil
color
colorless to light yellow
storage temp.
−20°C
SMILES string
CC(C1=O)=C[C@]2([H])[C@]1(O)CC(COC(C)=O)=C[C@]3([H])[C@]2(O)[C@H](C)C[C@]4(OC(CC5=CC=CC=C5)=O)[C@H]3C4(C)C
InChI
1S/C30H36O7/c1-17-11-23-28(34,26(17)33)15-21(16-36-19(3)31)12-22-25-27(4,5)29(25,14-18(2)30(22,23)35)37-24(32)13-20-9-7-6-8-10-20/h6-12,18,22-23,25,34-35H,13-16H2,1-5H3/t18-,22+,23-,25-,28-,29+,30-/m1/s1
InChI key
MEDVHSNRBPAIPU-XMOZQXTISA-N
Application
- to treat transfected HT-22 cells to promote phosphorylation of p66Shc
- to treat HT-22 and B12 cells to study its effects on cell viability
- to treat neurons and measure primary cortical neurons using the 2,5-diphenyl-2H-tetrazolium bromide (MTT) assay
Biochem/physiol Actions
Features and Benefits
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 1 Inhalation - Acute Tox. 2 Dermal - Acute Tox. 2 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Protein kinase C (PKC) is an AGC kinase that phosphorylates serine and threonine residues in many target proteins.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service