Skip to Content
Merck
All Photos(1)

Key Documents

D7808

Sigma-Aldrich

PCR Low Ladder Set

marker set for electrophoresis of PCR fragments

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

grade

Molecular Biology
for molecular biology

form

liquid

usage

50 uses

suitability

suitable for electrophoresis (DNA)

storage temp.

2-8°C

General description

Sigma′s PCR PCR Low Ladder Set contains one vial each of the PCR 100 bp Low Marker and the PCR 20 bp Low Marker. When mixed together, the ladder appears with bands every 20 bp, with every 5th band more intense. About two ul of the mixed ladder should be diluted in gel loading buffer and then loaded in a single lane on an agarose or polyacrylamide gel.

Components

Sigma′s PCR Low Ladder Set is provided as three separate tubes:
1 vial (25 ug) PCR 100 bp Low Marker is provided in a solution of 10 mM Tris-HCl (pH 8.0), with 1.0 mM EDTA.
1 vial (25 ug) PCR 20 bp Low Marker is provided in a solution of 10 mM Tris-HCl (pH 8.0), with 1.0 mM EDTA.
1 vial 6x loading buffer (250 ul volume) containing 15% Ficoll, 0.06% bromophenol blue, 0.06% xylene cyanol FF, and 30 mM EDTA.
1 additional empty tube is provided for storing the mixture.

Other Notes

For optimal resolution, the recommended agarose gel concentration is 2.5%.

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Donatella Panatto et al.
Human vaccines & immunotherapeutics, 11(1), 270-276 (2014-12-09)
Infections caused by Chlamydia trachomatis (Ct) and human papillomavirus (HPV) are the two main sexually transmitted infections; however, epidemiological data on Ct prevalence and Ct/HPV co-infection in Italy are scant. This study aimed at estimating the prevalence of Ct infection

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service