Skip to Content
Merck
All Photos(1)

Key Documents

D6288

Sigma-Aldrich

6α,9α-Difluoroprednisolone 21-acetate 17-butyrate

≥98%

Synonym(s):

Difluprednate

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
$140.00

$140.00


Estimated to ship on25 May 2025

New, lower price on this item!

Request a Bulk Order

Select a Size

Change View
50 MG
$140.00

About This Item

Empirical Formula (Hill Notation):
C27H34F2O7
CAS Number:
Molecular Weight:
508.55
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

$140.00


Estimated to ship on25 May 2025

New, lower price on this item!

Request a Bulk Order

sterility

non-sterile

Assay

≥98%

form

powder

solubility

chloroform: 50 mg/mL, clear, colorless

shipped in

ambient

storage temp.

2-8°C

SMILES string

CCCC(=O)O[C@@]1(CC[C@H]2[C@@H]3C[C@H](F)C4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]12C)C(=O)COC(C)=O

InChI

1S/C27H34F2O7/c1-5-6-23(34)36-26(22(33)14-35-15(2)30)10-8-17-18-12-20(28)19-11-16(31)7-9-24(19,3)27(18,29)21(32)13-25(17,26)4/h7,9,11,17-18,20-21,32H,5-6,8,10,12-14H2,1-4H3/t17-,18-,20-,21-,24-,25-,26-,27-/m0/s1

InChI key

WYQPLTPSGFELIB-JTQPXKBDSA-N

Gene Information

human ... NR3C1(2908)

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eric J Sigler et al.
Retina (Philadelphia, Pa.), 34(2), 237-246 (2013-07-25)
To describe the clinical and histopathologic characteristics of inflammatory deposits occurring within intermediate duration postoperative perfluoro-n-octane (PFO) for inferior retinal detachment repair. Prospective interventional case series of consecutive patients with inferior retinal detachment treated with intermediate duration postoperative vitreous cavity
Yixuan Dong et al.
Journal of pharmaceutical sciences, 108(6), 2002-2011 (2019-01-15)
Pharmaceutical emulsions contain multiple components, such as micellar, aqueous, and oil phases, leading to complex drug transfer and equilibrium phenomena. These complex components present challenges for the bioequivalence assessment of the drug products. The objective of the study was to
Shin-ichi Yasueda et al.
Journal of pharmaceutical and biomedical analysis, 30(6), 1735-1742 (2002-12-18)
A specific and sensitive method for the determination for difluprednate (DFBA) and its metabolite (deacetylated DFBA, DFB) in aqueous humor was developed. DFBA and DFB were initially absorbed on a Pinkerton-type column, then analyzed by high-performance liquid chromatography using a
Mahmoud Tawfik KhalafAllah et al.
BMJ open, 9(11), e026752-e026752 (2019-11-05)
Topical steroids are the cornerstone in controlling the inflammation after cataract surgery. Prednisolone acetate and difluprednate are the two main products for this purpose. However, it is unclear which one should be used in terms of effectiveness and safety. Systematic
Eric D Donnenfeld et al.
Journal of cataract and refractive surgery, 47(2), 158-164 (2020-09-15)
To evaluate the safety and efficacy of a nepafenac punctal plug delivery system (N-PPDS) after cataract surgery. Three U.S. clinical sites. Prospective, multicenter, randomized (2:1), parallel-arm, double-masked, placebo-controlled, phase II pilot study. Fifty-six subjects (aged older than 22 years) with

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service