Skip to Content
Merck
All Photos(1)

Key Documents

D103

Sigma-Aldrich

(±)-2,3-Dichloro-α-methylbenzylamine hydrochloride

solid

Synonym(s):

DCMB, LY-78335

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H9Cl2N · HCl
CAS Number:
Molecular Weight:
226.53
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

form

solid

color

white

solubility

H2O: soluble (Solutions may be stored for 1-2 days at 4 °C.)
ethanol: soluble (Solutions may be stored for 1-2 days at 4 °C.)

SMILES string

Cl[H].CC(N)c1cccc(Cl)c1Cl

Gene Information

human ... PNMT(5409)

Biochem/physiol Actions

PNMT inhibitor.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S P Kalra et al.
Endocrinology, 111(4), 1403-1405 (1982-10-01)
Administration of the opiate receptor antagonist, naloxone, evoked rapid rises in plasma LH levels in estrogen, progesterone-primed ovariectomized rats. Pretreatment with a peripherally acting epinephrine (EPI) synthesis inhibitor, SK&F 29661, failed to influence the naloxone-induced LH release. However, two centrally
T A Glauser et al.
Xenobiotica; the fate of foreign compounds in biological systems, 23(6), 657-669 (1993-06-01)
1. Thiol methyltransferase (TMT) is a microsomal enzyme catalyzing the S-methylation of aliphatic sulphydryl drugs and xenobiotics. Studies of the functional significance of S-methylation catalysed by TMT have been hampered by lack of a potent, relatively specific, non-toxic inhibitor of
R W Fuller
Annual review of pharmacology and toxicology, 22, 31-55 (1982-01-01)
Neurons that contain PNMT, the epinephrine-forming enzyme, have their cell bodies in brain stem regions in rat brain and send projections mainly into other brain stem areas, hypothalamus, and spinal cord. These neurons can be affected pharmacologically by various kinds
G Engberg et al.
Brain research, 223(1), 49-58 (1981-10-26)
The functional significance of the morphologically identified adrenaline (A)-mediated input to the noradrenergic nucleus locus coeruleus (LC) was pharmacologically analyzed. By means of single unit recording techniques the LC neurons in the rat brain were studied following administration of SK&F
N Y Liang et al.
The Journal of pharmacology and experimental therapeutics, 223(2), 382-387 (1982-11-01)
Intraperitoneal injections of the phenylethanolamine N-methyltransferase inhibitors 2-cyclohexyl-2-hydroxyethylamine, 1-aminomethylcycloundecanol, 7,8-dichloro-1,2,3,4-tetrahydroisoquinoline and 2,3-dichloro-alpha-methylbenzylamine all lowered blood pressures in spontaneously hypertensive rats. The drugs did not affect the blood pressures of age-paired normotensive Wistar Kyoto rats or did the drugs alter the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service