Skip to Content
Merck
All Photos(1)

Key Documents

D026

Sigma-Aldrich

Pargyline hydrochloride

Synonym(s):

N-Methyl-N-(2-propynyl)benzylamine hydrochloride, N-Methyl-N-propargylbenzylamine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2N(CH3)CH2C≡CH·HCl
CAS Number:
Molecular Weight:
195.69
Beilstein:
3699487
EC Number:
MDL number:
UNSPSC Code:
12352204

mp

160-163 °C (lit.)

storage temp.

−20°C

SMILES string

Cl[H].CN(CC#C)Cc1ccccc1

InChI

1S/C11H13N.ClH/c1-3-9-12(2)10-11-7-5-4-6-8-11;/h1,4-8H,9-10H2,2H3;1H

InChI key

BCXCABRDBBWWGY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Selective MAO-B inhibitor.

replaced by

Product No.
Description
Pricing

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service