Skip to Content
Merck
All Photos(7)

Key Documents

CWA-1067

Sigma-Aldrich

Anti-Human CD56 (MY31) ColorWheel® Dye-Ready mAb

greener alternative

for use with ColorWheel® Dyes (Required, sold separately)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Product CWA-1067 is not currently sold in your country. Contact Technical Service

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

MY31, monoclonal

product line

ColorWheel®

form

lyophilized

mol wt

calculated mol wt 94.57 kDa

species reactivity

human

species reactivity (predicted by homology)

nonhuman primates

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

technique(s)

flow cytometry: suitable

isotype

IgG1κ

epitope sequence

Unknown

Protein ID accession no.

UniProt accession no.

compatibility

for use with ColorWheel® Dyes (Required, sold separately)

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... NCAM1(4684)

General description

ColorWheel® technology is a novel and proprietary method of creating your own antibody and dye combinations for use in flow cytometry. By incubating any ColorWheel® antibody with any ColorWheel® dye, researchers can quickly and simply produce primary conjugated antibodies for use in single color or multicolor flow cytometry analysis. Each ColorWheel® antibody and dye is lyophilized for long long-term storage, allowing for simplicity and flexibility without compromise. ColorWheel® technology requires both ColorWheel® antibody and ColorWheel® dye for Flow Cytometry application.
We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This product is Preservative-free, lyophilized product for enhanced stability and allow for ambient shipping and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.

Specificity

Clone MY31 is a mouse monoclonal antibody that detects Neural cell adhesion molecule 1 (CD56).

Immunogen

KG1a leukemia cells

Application

Quality Control Testing:

Evaluated by Flow Cytometry in Human peripheral blood mononuclear cells (PBMC).

Flow Cytometry Analysis (FC): Staining of one million Human peripheral blood mononuclear cells (PBMC) was performed using 5 μL of a 1:1 mixture of Cat. No. CWA-1067, Anti-Human CD56 (MY31) ColorWheel® Dye-Ready mAb and Cat. No. CWD-PE ColorWheel® Antibody-Ready Phycoerythrin (PE) Dye or an equivalent amount of PE-conjugated Mouse IgG1 isotype control.

Evaluated by Flow Cytometry in Human NK cells.

Staining of one million Human NK cells was performed using 5 μL of a 1:1 mixture of Cat. No. CWA-1067, Anti-Human CD56 (MY31) ColorWheel® Dye-Ready mAb and Cat. No. CWD-PE ColorWheel® Antibody-Ready Phycoerythrin (PE) Dye (yellow histogram) or an equivalent amount of PE-conjugated Mouse IgG1 isotype control.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Compatibility

Requires ColorWheel® Dye (Sold Separately)

Target description

Neural cell adhesion molecule 1 (UniProt: P13591; also known as N-CAM-1, NCAM-1, CD56) is encoded by the NCAM1 (also known as NCAM) gene (Gene ID: 4684) in human. CD56 is a member of the immunoglobulin superfamily that is engaged in both so-called homophilic and heterophilic interactions. It is a single-pass type I membrane glycoprotein that is synthesized with a signal peptide (aa 1-19), which is subsequently cleaved off to generate the mature form that contains an extracellular domain (aa 20-718), a transmembrane domain (aa 719-739), and a cytoplasmic domain (aa 740-858). It also contains five Ig-like C2-type domains and two fibronectin type-III domains. CD56 is expressed by neurons, glia, skeletal muscle, and natural killer (NK) cells. It is involved in neuron-neuron adhesion, neurite fasciculation, and outgrowth of neurite. It is considered as the archetypal phenotypic marker for natural killer cells, but is also expressed in many other immune cells, including ab T cells, g T cells, dendritic cells, and monocytes. It is also shown to play a role in the fungus mediated NK cell activation and blocking of CD56 surface receptor reduced fungal mediated NK cell activation and reduced cytokine secretion. (Ref.: Ziegler, S., et al. (2017). Sci. Rep. 7; Article 6138; Van Acker, HH., et al. (2017). Front. Immunol. 8; Article 892).

Physical form

Lyophilized from PBS containing D-Mannitol and Sucrose, normal appearance is a dried pellet. Reconstituted antibody solution is stable and functional as assessed by functional testing. Contains no biocide or preservatives, such as azide.

Reconstitution

1.0 mg/mL after reconstitution at 25 μL in PBS. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C. Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial. Reconstitute each vial by adding 25 μL of PBS. Protect from light.

Legal Information

ColorWheel is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

required but not provided

Product No.
Description
Pricing

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service