Skip to Content
Merck
All Photos(1)

Key Documents

CLS8505

Corning® screw cap

polypropylene, with o-ring on mat, clear with cover

Synonym(s):

Corning screw cap

Sign Into View Organizational & Contract Pricing

Select a Size

480 EA
$833.00
10 X 480 EA
$7,380.00

$833.00


Estimated to ship on13 May 2025



Select a Size

Change View
480 EA
$833.00
10 X 480 EA
$7,380.00

About This Item

UNSPSC Code:
41121800
NACRES:
NB.15

$833.00


Estimated to ship on13 May 2025


material

polypropylene

sterility

non-sterile

manufacturer/tradename

Corning

size

96 wells

color

clear

Looking for similar products? Visit Product Comparison Guide

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service