CLS6775
Corning® LSE™ vortex mixers
vortex mixer w/ standard tube head, 120 V, 1/cs
Synonym(s):
laboratory mixer shaker, laboratory mixers, vortexers
About This Item
Recommended Products
AC/DC input
120 V AC
feature
CE compliant
CSA approved
packaging
case of 1
pack of 1
manufacturer/tradename
Corning 6775
parameter
0-3400 rpm speed
L × W × H
14 cm × 16 cm × 13 cm
weight
2.2 kg
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
- Variable speed for gentle to vigorous mixing
- Optimized counter balance for minimal vibration
- Lightweight and portable
- Includes cup head for all standard microtubes and centrifuge tubes
- Mix a variety of tubes and plates with optional accessory heads
- Touch and continuous mix modes
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service