Key Documents
CLS6406500FO
PYREX® replacement Squibb separatory funnel w/o valve and stopper
pack of 4 ea
Synonym(s):
equipment replacement accessory
Select a Size
4 EA
$573.00
$573.00
Check Cart for Availability
Select a Size
Change View
4 EA
$573.00
About This Item
UNSPSC Code:
41121809
$573.00
Check Cart for Availability
packaging
pack of 4 ea
manufacturer/tradename
Corning 6406-500FO
capacity
500 mL
volume
500 mL
Related Categories
General description
These economy Squibb separatory funnel replacements do not have a Polytetrafluorethylene (PTFE) valve assembly or polyethylene stopper.
Legal Information
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)