CLS56551L
PYREX® EZ Access™ wide mouth volumetric flask
capacity 1 L
Synonym(s):
volumetric glassware
Select a Size
$221.00
Select a Size
About This Item
$221.00
Recommended Products
material
PTFE liner
cap (polybutylene terephthalate)
flask (class A glass)
flask (flat bottom, threaded)
description
heavy duty, with GL32 screw cap
feature
class A
To Contain
packaging
pack of 1 ea
manufacturer/tradename
Corning 5655-1L
H × O.D.
315 mm × 132 mm
capacity
1 L
fitting
GL 32 thread
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service