Documents
CLS480140
Corning® LSE Adapters
for 10/15 mL tubes in 50 mL rotor, 6/cs
Synonym(s):
centrifuge rotor tube adapters, centrifuge tube adapters
About This Item
UNSPSC Code:
41121815
NACRES:
SB.56
packaging
case of 6
pack of 6
manufacturer/tradename
Corning 480140
suitability
suitable for (for use with Corning LSE compact centrifuge)
Related Categories
General description
These are optional adapters for Corning LSE compact centrifuge rotors: CLS480136, CLS480137, CLS480138; CLS480139
Legal Information
Corning is a registered trademark of Corning, Inc.
LSE is a trademark of Corning, Inc.