Skip to Content
Merck
All Photos(1)

Key Documents

CLS4714

Corning® universal fit hinged racked pipet tips

100-1000 μL pipet tips, blue, sterile, 100tips/rack, 10 racks/case, 1000 tips/case

Synonym(s):

pipet tips, pipette tips

Sign Into View Organizational & Contract Pricing

Select a Size

1000 EA
$89.50

$89.50


Check Cart for Availability


Select a Size

Change View
1000 EA
$89.50

About This Item

UNSPSC Code:
41121600
NACRES:
NB.01
volume:
100—1000 μL
sterility:
sterile
feature:
DNase free
RNase free
barrier: no
disposable
endotoxin free
filter: no
graduation: no
insert racks without hinged lid
packaging:
rack of 100
manufacturer/tradename:
Corning 4714

$89.50


Check Cart for Availability

material

blue tip
round polypropylene tip

sterility

sterile

feature

DNase free
RNase free
barrier: no
disposable
endotoxin free
filter: no
graduation: no
insert racks without hinged lid

packaging

rack of 100

manufacturer/tradename

Corning 4714

volume range

100-1000 μL

volume

100—1000 μL

Looking for similar products? Visit Product Comparison Guide

General description

Corning® universal fit hinged rack pipet tips are designed to assure a reliable fit with all major brand pipettors. Tips are available in two sizes: 200 μL and 1000 μL. The hinged rack design allows easy rack stacking. The hinged closure allows steam penetration during autoclaving. Racks can be reused. Simply remove the tip insert in the rack and replace with the Corning tip inserts for hinged racks.

  • For use with 1-200 μL and 100-1000 μL single and multichannel pipettors
  • 200 μL racks have 96 tips per rack; 1000 μL racks have 100 tips per rack
  • Exceptional durability - all Corning tip racks stand up to the demands of use with multichannel pipettors and withstand repeated autoclaving cycles
  • Certified RNase-/DNase-free and nonpyrogenic
  • Available sterile and nonsterile
  • Tips are not graduated

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service