CLS4710
Corning® universal fit hinged racked pipet tips
1-200 μL pipet tips, yellow, non-sterile, 96 tips/rack, 10 racks/case, 960 tips/case
Synonym(s):
pipet tips, pipette tips
Select a Size
$50.70
Select a Size
About This Item
RNase free
barrier: no
disposable
endotoxin free
filter: no
graduation: no
insert racks without hinged lid
$50.70
Recommended Products
material
round polypropylene tip
yellow tip
sterility
non-sterile
feature
DNase free
RNase free
barrier: no
disposable
endotoxin free
filter: no
graduation: no
insert racks without hinged lid
packaging
rack of 96
manufacturer/tradename
Corning 4710
volume range
1-200 μL
volume
1—200 μL
Looking for similar products? Visit Product Comparison Guide
General description
- For use with 1-200 μL and 100-1000 μL single and multichannel pipettors
- 200 μL racks have 96 tips per rack; 1000 μL racks have 100 tips per rack
- Exceptional durability - all Corning tip racks stand up to the demands of use with multichannel pipettors and withstand repeated autoclaving cycles
- Certified RNase-/DNase-free and nonpyrogenic
- Available sterile and nonsterile
- Tips are not graduated
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service