CLS4572
Corning® 1536 well plates, cyclic olefin co-polymer
white, NBS, non-sterile
Synonym(s):
1536 multiwell plate
Select a Size
$3,300.00
Estimated to ship on11 May 2025
Select a Size
About This Item
$3,300.00
Estimated to ship on11 May 2025
material
white
sterility
non-sterile
feature
NBS
packaging
pack of 10 ea
case of 50 ea
manufacturer/tradename
Corning 4572
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Key Features
- Cyclic Olefin Copolymer (COC) material
- Flat top for improved sealing
- 1536 Well Hi Base solid color microplates (black or white)
- Variety of treatment options
- tissue culture treated
- Corning CellBIND® Surface
- Nonbinding Surface (NBS)
- Not treated
- Bar coded (one label on both long sides)
- Custom bar codes available for compatibility with the Kalypsys System
- Low autofluorescence
- Broad chemical resistance including DMSO and alcohol, which allows for assay step reduction
- High mechanical stability
- Optimized for flatness and uniformity
- Low birefringence
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service