Key Documents
CLS451536L
Corning® direct overhead drive assembly for ProCulture® spinner flasks
paddle assembly for 36L Proculture spinner flask w/ direct drive overhead motor, 1/cs
Synonym(s):
Corning container
Select a Size
1 EA
$346.00
$346.00
Check Cart for Availability
Select a Size
Change View
1 EA
$346.00
About This Item
UNSPSC Code:
41122104
NACRES:
NB.43
$346.00
Check Cart for Availability
sterility
non-sterile
packaging
case of 1
pack of 1
manufacturer/tradename
Corning 4515-36L
General description
Direct drive paddle assembly for use with direct drive overhead motors on series 4510 and 4512, 36L Proculture® Spinner Flasks. The paddle assemblies can be used with either 100 mm or 120 mm direct drive center cap assemblies.
Legal Information
Corning is a registered trademark of Corning, Inc.
ProCulture is a registered trademark of SP Industries
Choose from one of the most recent versions:
Certificates of Analysis (COA)