CLS440812
Corning® SS2I Stirrer
Single position, Slow Speed, capacity 10 L, AC/DC input 120 V AC
Synonym(s):
Corning stirrer accessory
About This Item
Recommended Products
material
platform (glass filled resin)
feature
Single position, Slow Speed
measuring range
10-150 rpm
capacity
10 L
vessel diam.
≤280 mm
weight
2.6 kg
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service