Skip to Content
Merck
All Photos(1)

Key Documents

CLS430599

Corning® tissue-culture treated culture dishes

D × H 150 mm × 25 mm

Synonym(s):

petri dish

Sign Into View Organizational & Contract Pricing

Select a Size

5 EA
$45.20
60 EA
$105.00

$45.20


Available to ship on28 April 2025Details



Select a Size

Change View
5 EA
$45.20
60 EA
$105.00

About This Item

UNSPSC Code:
41121812
NACRES:
NA.75

$45.20


Available to ship on28 April 2025Details


material

polystyrene
round

sterility

sterile

packaging

sleeve of 5

manufacturer/tradename

Corning 430599

D × H

150 mm × 25 mm

surface area

148 cm2

Looking for similar products? Visit Product Comparison Guide

General description

["Corning"]Tissue Culture (TC)-treated products are surface modified by corona discharge or gas plasma treatment which leaves the surface hydrophilic and negatively charged with the addition of media.

Application

["Corning"] tissue-culture-treated culture dishes are suitable for everyday attachment cell culture needs.[1][2]

Features and Benefits

  • Manufactured from optically clear virgin polystyrene
  • Treated for optimal cell attachment
  • Sterilized by gamma radiation and certified nonpyrogenic
  • Have stacking beads to aid in handling
  • Lid is supplied with vents to provide consistent gas exchange

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Paul T Toran et al.
Experimental hematology, 86, 28-42 (2020-05-31)
As important vectors for ectopic protein expression, gene silencing, and progenitor cell barcoding, lentiviruses continue to emerge as versatile research and clinical tools. For studies employing cell types that are relatively resistant to transduction, high-titer lentivirus preparations with low cytotoxicity
Min Shen et al.
Redox biology, 47, 102151-102151 (2021-10-05)
Ferroptosis is a recently identified non-apoptotic form of cell death characterized by iron-dependent lipid peroxidation. However, the underlying exact mechanisms remain poorly understood. Here, we report that the total levels of N6-methyladenosine (m6A) modification are evidently increased upon exposure to
Zili Zhang et al.
Redox biology, 36, 101619-101619 (2020-08-31)
Ferroptosis is a recently discovered form of programmed cell death, but its regulatory mechanisms are not fully understood. In the current study, we reported that the BRD7-P53-SLC25A28 axis played a crucial role in regulating ferroptosis in hepatic stellate cells (HSCs).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service