Skip to Content
Merck
All Photos(1)

Key Documents

CLS4124

Corning® DeckWorks standard pipet tips

50 - 1250μL extended length pipet tips, graduated, hinged racks, natural, sterile, polypropylene, 96 tips/rack, 8 racks/pack, 4 packs/case

Synonym(s):

pipet tips, pipette tips

Sign Into View Organizational & Contract Pricing

Select a Size

768 EA
$169.00
4 X 768 EA
$423.00

$169.00


Check Cart for Availability


Select a Size

Change View
768 EA
$169.00
4 X 768 EA
$423.00

About This Item

UNSPSC Code:
41121600
NACRES:
NB.02
volume:
50—1250 μL
sterility:
sterile
feature:
DNase free
RNase free
barrier: no
disposable
endotoxin free
filter: no
graduated
Pipet tip: round
packaging:
rack of 96
manufacturer/tradename:
Corning 4124

$169.00


Check Cart for Availability

material

natural tip
polypropylene

sterility

sterile

feature

DNase free
RNase free
barrier: no
disposable
endotoxin free
filter: no
graduated
Pipet tip: round

packaging

rack of 96

manufacturer/tradename

Corning 4124

volume range

50-1250 μL

volume

50—1250 μL

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Corning® DeckWorks Pipet Tips

Bulk Pipet Tips
Corning DeckWorks bulk pipet tips are compatible with DeckWorks racks and packaged in resealable polyethylene bags. Pipet tip sizes are as follows: 10 uL, 200 uL and 1000 uL.

Universal Fit Pipet Tips
Corning universal fit tips in hinged DeckWorks racks are available in 10 uL, 200 uL, 300 uL and 1000 uL sizes. The 10 uL, 300 uL and 1000 uL tips are extended length and allow for improved access into micro (1.5 mL) and standard conical bottom (15/50 mL) centrifuge tubes.

Reload Systems
Robust 96-tip racks are designed for repeated steam autoclave cycles and can be reused by removing the empty grey tip deck and replacing it with a new reload deck of 96 tips from the DeckWorks reload system. Corning racked 300 uL DeckWorks pipet tips are designed for all popular 300 uL multi-channel instruments.

This system is designed to allow you to maximize storage space and help reduce the production and waste of plastic materials. Both the hinged racks and reload decks within this system are manufactured with recycled plastic. The pipet tips are made in a controlled molding environment to ensure accuracy and precision across all tip volumes.

Medical grade resins and production excellence allow for full certification as:

  • Nonpyrogenic
  • Human DNA-free
  • RNase-/DNase-free

Legal Information

Corning is a registered trademark of Corning, Inc.
DeckWorks is a trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service