Skip to Content
Merck
All Photos(1)

Key Documents

CLS4112

Corning® DeckWorks standard pipet tips

1 - 200μL pipet tips, graduated, yellow, non-sterile, polypropylene, 1000 tips/pack, 10,000 tips/case

Synonym(s):

pipet tips, pipette tips

Sign Into View Organizational & Contract Pricing

Select a Size

1000 EA
$67.50
10 X 1000 EA
$209.00

$67.50


Check Cart for Availability


Select a Size

Change View
1000 EA
$67.50
10 X 1000 EA
$209.00

About This Item

UNSPSC Code:
41121600
NACRES:
NB.01
volume:
1—200 μL
sterility:
non-sterile
feature:
DNase free
RNase free
barrier: no
disposable
endotoxin free
filter: no
graduated
Pipet tip: round
packaging:
pack of 10000
manufacturer/tradename:
Corning 4112

$67.50


Check Cart for Availability

material

polypropylene
yellow tip

sterility

non-sterile

feature

DNase free
RNase free
barrier: no
disposable
endotoxin free
filter: no
graduated
Pipet tip: round

packaging

pack of 10000

manufacturer/tradename

Corning 4112

volume range

1-200 μL

volume

1—200 μL

Looking for similar products? Visit Product Comparison Guide

General description

Corning® DeckWorks Pipet Tips

Bulk Pipet Tips
Corning DeckWorks bulk pipet tips are compatible with DeckWorks racks and packaged in resealable polyethylene bags. Pipet tip sizes are as follows: 10 uL, 200 uL and 1000 uL.

Universal Fit Pipet Tips
Corning universal fit tips in hinged DeckWorks racks are available in 10 uL, 200 uL, 300 uL and 1000 uL sizes. The 10 uL, 300 uL and 1000 uL tips are extended length and allow for improved access into micro (1.5 mL) and standard conical bottom (15/50 mL) centrifuge tubes.

Reload Systems
Robust 96-tip racks are designed for repeated steam autoclave cycles and can be reused by removing the empty grey tip deck and replacing it with a new reload deck of 96 tips from the DeckWorks reload system. Corning racked 300 uL DeckWorks pipet tips are designed for all popular 300 uL multi-channel instruments.

This system is designed to allow you to maximize storage space and help reduce the production and waste of plastic materials. Both the hinged racks and reload decks within this system are manufactured with recycled plastic. The pipet tips are made in a controlled molding environment to ensure accuracy and precision across all tip volumes.

Medical grade resins and production excellence allow for full certification as:

  • Nonpyrogenic
  • Human DNA-free
  • RNase-/DNase-free

Legal Information

Corning is a registered trademark of Corning, Inc.
DeckWorks is a trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service