Skip to Content
Merck
All Photos(1)

Key Documents

CLS4074

Corning® Lambda® plus single channel pipettor

volume 20-200 μL

Synonym(s):

single-channel pipettor

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$393.00

$393.00


Available to ship on14 April 2025Details



Select a Size

Change View
1 EA
$393.00

About This Item

UNSPSC Code:
41121500
NACRES:
NB.01

$393.00


Available to ship on14 April 2025Details


feature

autoclavable
single-channel

manufacturer/tradename

Corning 4074

volume measuring range

20-200 μL

volume

200 μL

application(s)

PFAS testing

Looking for similar products? Visit Product Comparison Guide

Related Categories

Legal Information

Corning is a registered trademark of Corning, Inc.
Lambda is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

FAS, or 'forever chemicals,' persist in the environment and pose risks to human health. Discover our environmental monitoring tools for PFAS quantification, aiding researchers, regulators, and labs in testing for PFAS.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service