Skip to Content
Merck
All Photos(1)

Key Documents

CLS3884

Corning® 96 Well Half-Area Microplate

flat bottom clear, white polystyrene, (NBS), bag of 25, non-sterile, lid: no

Synonym(s):

96 multiwell plate

Sign Into View Organizational & Contract Pricing

Select a Size

100 EA
$2,030.00

$2,030.00


Check Cart for Availability


Select a Size

Change View
100 EA
$2,030.00

About This Item

UNSPSC Code:
41121800
NACRES:
NB.15
material:
clear bottom
flat bottom clear
white polystyrene
size:
96 wells

$2,030.00


Check Cart for Availability

material

clear bottom
flat bottom clear
white polystyrene

sterility

non-sterile

feature

lid: no
skirt
(NBS)

packaging

bag of 25
pkg of 25 inner package

manufacturer/tradename

Corning 3884

size

96 wells

well D

10.7 mm

well volume

190 μL

wells

96 wells , half area

working volume

25-125 μL

suitability

suitable for (cell cuture)

binding type

NBS (non-binding surface)

Looking for similar products? Visit Product Comparison Guide

General description

Unique half area plates can help save on rare and expensive reagents by reducing the amount of reagent needed per well, while still being readable by most standard plate readers.

Legal Information

Corning is a registered trademark of Corning, Inc.
NBS is a trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service