Skip to Content
Merck
All Photos(1)

Key Documents

CLS3797

Corning® 96 Well EIA/RIA Assay Microplate

round bottom, clear polystyrene, medium binding surface, bag of 25, non-sterile, lid: no

Synonym(s):

96 multiwell plate, 96 well microplate, 96 well microtiter plate, 96 well plate, EIA/RIA plate, immunoassay plate

Sign Into View Organizational & Contract Pricing

Select a Size

100 EA
$209.00

$209.00


Available to ship on14 April 2025Details



Select a Size

Change View
100 EA
$209.00

About This Item

UNSPSC Code:
41121800
NACRES:
NB.13
material:
clear bottom
clear polystyrene
polystyrene
round bottom
round wells
size:
96 wells , 96 well plate

$209.00


Available to ship on14 April 2025Details


material

clear bottom
clear polystyrene
polystyrene
round bottom
round wells

sterility

non-sterile

feature

lid: no
skirt
plate format: 96 well standard

packaging

case of 100
bag of 25

manufacturer/tradename

Corning 3797

size

96 wells , 96 well plate

well volume

330-360 μL , Note: CLS3366 and CLS3797 have well volume capacities of 330 uL

well working volume

75-200 μL

color

clear plate

suitability

suitable for (EIA/RIA; immunoassays)

binding type

medium binding surface

Looking for similar products? Visit Product Comparison Guide

General description

  • Not treated (or medium binding) polystyrene surface is hydrophobic in nature and binds biomolecules through passive interactions
  • Suitable primarily for the immobilization of large molecules, such as antibodies, that have large hydrophobic regions that can interact with the surface
  • Binding capacity of approximately 250 ng of mouse IgG/cm2
  • EIA/RIA plates are tested on lot-to-lot basis and the certification results for each lot are made available upon request
  • Recommended working volume of up to 75 to 200 μL
  • Conforms to standard microplate footprint and dimensions
  • Without lids, lids supplied separately
These 96 well EIA/RIA plates are designed for in vitro diagnostic immunoassays. All EIA/RIA plates are manufactured from special medical-grade polystyrene for uniform binding, high optical clarity and low background absorption. In addition, the Easy-Wash products are part of the Certified Surface Chemistry Programme, which guarantees protein binding performance to be within QA release specifications.

Legal Information

Corning is a registered trademark of Corning, Inc.
Easy-Wash is a trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service